Skip to Content
Merck
All Photos(1)

Key Documents

SML2486

Sigma-Aldrich

Peramivir trihydrate

≥98% (HPLC)

Synonym(s):

(1S,2S,3R,4R)-3-[(1S)-1-(Acetylamino)-2-ethylbutyl]-4-[(aminoiminomethyl)amino]-2-hydroxycyclopentanecarboxylic acid trihydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H28N4O4 · 3H2O
CAS Number:
Molecular Weight:
382.45
UNSPSC Code:
51111800
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D -125 to -145°, c = 0.1 in H2O

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

N([C@H]1[C@@H]([C@@H]([C@H](C1)C(=O)O)O)[C@@H](NC(=O)C)C(CC)CC)C(=N)N.O.O.O

InChI

1S/C15H28N4O4.3H2O/c1-4-8(5-2)12(18-7(3)20)11-10(19-15(16)17)6-9(13(11)21)14(22)23;;;/h8-13,21H,4-6H2,1-3H3,(H,18,20)(H,22,23)(H4,16,17,19);3*1H2/t9-,10+,11+,12-,13+;;;/m0.../s1

InChI key

RFUCJKFZFXNIGB-ZBBHRWOZSA-N

Biochem/physiol Actions

Influenza viral neuraminidase inhibitor
Peramivir exhibits antiviral activity against type A and B influenza virus including pandemic influenza A/H1N1 virus. It also exhibits its antiviral effects on avian influenza viruses like H5N1 and H9N2 strains.
Peramivir is an Influenza viral neuraminidase inhibitor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masanori Kobayashi et al.
Antiviral research, 139, 41-48 (2016-12-26)
High morbidity and mortality associated with human cases of highly pathogenic avian influenza (HPAI) viruses, including H5N1 influenza virus, have been reported. The purpose of the present study was to evaluate the antiviral effects of peramivir against HPAI viruses. In
Zhengtu Li et al.
BMC infectious diseases, 16, 325-325 (2016-07-10)
China-made Peramivir, an anti-influenza neuraminidase inhibitor drug, is manufactured and widely used in China. Although effective if initiated within 48 h of the onset of symptoms, yet we observed that this drug shows an inconclusive efficacy if treatment is delayed in
Jung-Wan Yoo et al.
Journal of medical virology, 87(10), 1649-1655 (2015-05-07)
The clinical efficacy of peramivir in the treatment of severe seasonal influenza in critically ill patients admitted to an intensive care unit (ICU) is not well established. The aim of this study was to compare the clinical efficacy of peramivir
Makoto Kodama et al.
Antiviral research, 109, 110-115 (2014-07-06)
The purpose of this study was to investigate the relationship between pharmacokinetic (PK) parameters of intravenous (IV) peramivir and in vivo antiviral activity pharmacodynamic (PD) outcomes in a mouse model of influenza virus infection. Peramivir was administrated to mice in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service