Skip to Content
Merck
All Photos(1)

Key Documents

SML2418

Sigma-Aldrich

PS10

≥98% (HPLC)

Synonym(s):

2-[(2,4-Dihydroxyphenyl)sulfonyl]-2,3-dihydro-1H-isoindole-4,6-diol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H13NO6S
CAS Number:
Molecular Weight:
323.32
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=S(C1=C(O)C=C(O)C=C1)(N2CC(C(O)=CC(O)=C3)=C3C2)=O

InChI

1S/C14H13NO6S/c16-9-1-2-14(13(19)4-9)22(20,21)15-6-8-3-10(17)5-12(18)11(8)7-15/h1-5,16-19H,6-7H2

InChI key

SVUZJWAAXPEMKJ-UHFFFAOYSA-N

Biochem/physiol Actions

2-[(2,4-dihydroxyphenyl) sulfonyl]isoindoline-4,6-diol (PS10) can be considered as a therapeutic method to preserve glucose and lipid homeostasis in obesity and type 2 diabetes (T2D). It can induce pyruvate dehydrogenase complex (PDC) activity in liver and kidney. It is also considered as an effective PDK inhibitor to treat diabetic cardiomyopathy. In the normal myocardium, PS10 helps to enhance fractional carbohydrate oxidation.
PS10 is a potent and selective PDK (Pyruvate dehydrogenase kinase) inhibitor that improves glucose tolerance and lessens hepatic steatosis in diet-induced obese mice. PS10 binds to the ATP-binding pocket of PDKs.
potent and selective PDK (Pyruvate dehydrogenase kinase) inhibitor

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shih-Chia Tso et al.
The Journal of biological chemistry, 289(7), 4432-4443 (2013-12-21)
Pyruvate dehydrogenase kinase isoforms (PDKs 1-4) negatively regulate activity of the mitochondrial pyruvate dehydrogenase complex by reversible phosphorylation. PDK isoforms are up-regulated in obesity, diabetes, heart failure, and cancer and are potential therapeutic targets for these important human diseases. Here
Shih-Chia Tso et al.
Journal of medicinal chemistry, 60(3), 1142-1150 (2017-01-14)
Pyruvate dehydrogenase kinases 1-4 (PDK1-4) negatively control activity of the pyruvate dehydrogenase complex (PDC) and are up-regulated in obesity, diabetes, heart failure, and cancer. We reported earlier two novel pan-PDK inhibitors PS8 [4-((5-hydroxyisoindolin-2-yl)sulfonyl)benzene-1,3-diol] (1) and PS10 [2-((2,4-dihydroxyphenyl)sulfonyl)isoindoline-4,6-diol] (2) that targeted
Cheng-Yang Wu et al.
The Journal of biological chemistry, 293(25), 9604-9613 (2018-05-10)
The pyruvate dehydrogenase complex (PDC) is a key control point of energy metabolism and is subject to regulation by multiple mechanisms, including posttranslational phosphorylation by pyruvate dehydrogenase kinase (PDK). Pharmacological modulation of PDC activity could provide a new treatment for
Targeting hepatic pyruvate dehydrogenase kinases restores insulin signaling and mitigates ChREBP-mediated lipogenesis in diet-induced obese mice
Wu CY, et al.
Molecular Metabolism, 12, 12-24 (2018)
Mona Synnøve Bjune et al.
EBioMedicine, 91, 104569-104569 (2023-04-21)
The valine (branched-chain amino acid) metabolite 3-hydroxyisobutyrate (3-HIB), produced by 3-Hydroxyisobutyryl-CoA Hydrolase (HIBCH), is associated with insulin resistance and type 2 diabetes, but implicated tissues and cellular mechanisms are poorly understood. We hypothesized that HIBCH and 3-HIB regulate hepatic lipid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service