Skip to Content
Merck
All Photos(1)

Key Documents

SML2182

Sigma-Aldrich

Succinobucol

≥98% (HPLC)

Synonym(s):

AGI-1067, Butanedioic acid 1-[4-[[1-[[3,5-bis(1,1-dimethylethyl)-4-hydroxyphenyl]thio]-1-methylethyl]thio]-2,6-bis(1,1-dimethylethyl)phenyl]ester, Probucol monosuccinate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H52O5S2
CAS Number:
Molecular Weight:
616.91
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

OC1=C(C(C)(C)C)C=C(SC(C)(C)SC2=CC(C(C)(C)C)=C(OC(CCC(O)=O)=O)C(C(C)(C)C)=C2)C=C1C(C)(C)C

InChI

1S/C35H52O5S2/c1-31(2,3)23-17-21(18-24(29(23)39)32(4,5)6)41-35(13,14)42-22-19-25(33(7,8)9)30(26(20-22)34(10,11)12)40-28(38)16-15-27(36)37/h17-20,39H,15-16H2,1-14H3,(H,36,37)

InChI key

RKSMVPNZHBRNNS-UHFFFAOYSA-N

Biochem/physiol Actions

Orally active antioxidant; vascular protectant; antiatherosclerotic;
Succinobucol is a metabolically stable derivative of the antioxidant drug probucol that poses a distinct pharmacological from. Succinobucol is an orally active vascular protectant that exhibits antiatherosclerotic properties in multiple animal models and in humans. It inhibits the release of proinflammatory cytokines, TNF-a, IL-1b, and IL-6 in hPBMC activated by LPS.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stephanie A Houston et al.
Platelets, 28(3), 295-300 (2016-09-30)
Succinobucol is a phenolic antioxidant with anti-inflammatory and antiplatelet effects. Given the importance of oxidant stress in modulating platelet-platelet and platelet-vessel wall interactions, the aim of this study was to establish if antioxidant activity was responsible for the antiplatelet activity
Dirleise Colle et al.
Molecular neurobiology, 53(2), 1280-1295 (2015-01-27)
Succinobucol (succinyl ester of probucol) is a lipid-lowering compound with anti-inflammatory and antioxidant properties. Recent experimental evidence has highlighted the potential neuroprotective effects of succinobucol. In the present study, cultured neuroblastoma (SH-SY5Y) cells were used to investigate mechanisms mediating the
Charles Kunsch et al.
The Journal of pharmacology and experimental therapeutics, 308(3), 820-829 (2003-11-18)
Atherosclerosis is a disease of oxidative stress and inflammation. AGI-1067 [butanedioic acid, mono[4-[[1-[[3,5-bis(1,1-dimethylethyl)-4-,hydroxyphenyl]thio]-1-methylethyl]thio]-2,6-bis (1,1-dimethylethyl)phenyl] ester] is a metabolically stable derivative of, yet pharmacologically distinct from, the antioxidant drug probucol. It is a member of a novel class of orally active

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service