Skip to Content
Merck

SML2018

GSK4027

≥98% (HPLC)

Synonym(s):

4-Bromo-2-methyl-5-(((3R,5R)-1-methyl-5-phenylpiperidin-3-yl)-amino)pyridazin-3(2H)-one, 4-Bromo-2-methyl-5-[[(3R,5R)-1-methyl-5-phenyl-3-piperidinyl]amino]-3(2H)-pyridazinone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C17H21BrN4O
CAS Number:
Molecular Weight:
377.28
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

CN(C[C@H](C1)C2=CC=CC=C2)C[C@@H]1NC(C=NN(C3=O)C)=C3Br

InChI

1S/C17H21BrN4O/c1-21-10-13(12-6-4-3-5-7-12)8-14(11-21)20-15-9-19-22(2)17(23)16(15)18/h3-7,9,13-14,20H,8,10-11H2,1-2H3/t13-,14+/m0/s1

InChI key

VZAFGXCWAWRULT-UONOGXRCSA-N

Biochem/physiol Actions

GSK4027 is a cell penetrant, potent and selective inhibitor of PCAF/GCN5 bromodomain [p300/CBP-associated factor (PCAF)/general control nonderepressible 5 (GCN5) bromodomain].
cell penetrant, potent and selective inhibitor of PCAF/GCN5 bromodomain
It acts as a chemical probe for PCAF/GCN5.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library