Skip to Content

SML2004

Cangrelor tetrasodium salt

≥95% (HPLC), P2Y12 antagonist, powder

Synonym(s):

AR-C69931 tetrasodium salt, AR-C69931MX tetrasodium salt, N-[2-(methylthio)ethyl]-2-[(3,3,3-trifluoropropyl)thio]-, anhydride with P,P′-(dichloromethylene)bis[phosphonic acid]-5′-Adenylic acid tetrasodium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C17H21Cl2F3N5O12P3S2·Na4
CAS Number:
Molecular Weight:
864.29
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥95% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Cangrelor tetrasodium salt, ≥95% (HPLC)

assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

H2O: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

O[C@@H]1[C@H](O)[C@@H](COP(OP(C(Cl)(Cl)P(O[Na])(O[Na])=O)(O[Na])=O)(O[Na])=O)O[C@H]1N(C=N2)C3=C2C(NCCSC)=NC(SCCC(F)(F)F)=N3

InChI

1S/C17H25Cl2F3N5O12P3S2.4Na/c1-43-5-3-23-12-9-13(26-15(25-12)44-4-2-16(20,21)22)27(7-24-9)14-11(29)10(28)8(38-14)6-37-42(35,36)39-41(33,34)17(18,19)40(30,31)32;;;;/h7-8,10-11,14,28-29H,2-6H2,1H3,(H,33,34)(H,35,36)(H,23,25,26)(H2,30,31,32);;;;/q;4*+1/p-4/t8-,10-,11-,14-;;;;/m1..../s1

InChI key

COWWROCHWNGJHQ-OPKBHZIBSA-J

Application

Cangrelor has been used as a purinergic receptor P2Y12 antagonist to study its effects on neuromedin U receptor 2 (NMUR2)-coupled Gαi pathway during platelet aggregation.

Biochem/physiol Actions

Cangrelor is a potent and selective platelet P2Y12 antagonist. It is used as an intravenous antiplatelet drug to prevent formation of harmful blood clots in the coronary arteries.
Potent and selective platelet P2Y12 antagonist
Cangrelor is an analog of nonthienopyridine adenosine triphosphate. It has a therapeutic effect against reversible platelet inhibition.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library