Skip to Content
Merck

SML1759

CD1530

≥98% (HPLC)

Synonym(s):

4-(6-Hydroxy-7-tricyclo[3.3.1.13,7]dec-1-yl-2-naphthalenyl)-benzoic acid, CD-1530

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C27H26O3
CAS Number:
Molecular Weight:
398.49
NACRES:
NA.77
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

OC1=CC2=C(C=C(C3=CC=C(C(O)=O)C=C3)C=C2)C=C1C4(C[C@H](C5)C6)C[C@H]6C[C@H]5C4

InChI

1S/C27H26O3/c28-25-12-22-6-5-21(19-1-3-20(4-2-19)26(29)30)10-23(22)11-24(25)27-13-16-7-17(14-27)9-18(8-16)15-27/h1-6,10-12,16-18,28H,7-9,13-15H2,(H,29,30)

InChI key

VCQGNUWOMLYNNG-UHFFFAOYSA-N

Biochem/physiol Actions

CD1530 is a potent and selective agonist of retinoic acid receptor RARγ with Kd values of 150, 1,500, and 2,750 nM for RARγ, RARβ, and RARα receptors, respectively. In various studies, CD1530 has been shown to preserve human tendon stem cell characteristics, promote repair of injured skeletal muscle, and in combination with bexarotene to inhibit oral carcinogenesis.
CD1530 is a potent and selective agonist of retinoic acid receptor RARγ.


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library