Skip to Content
Merck
All Photos(1)

Documents

SML1651

Sigma-Aldrich

Iloprost

≥98% (HPLC)

Synonym(s):

(E)-(3aS, 4R, 5R, 6aS)-Hexahydro-5-hydroxy-4-[(E)-(3S,4RS)-3-hydroxy-4-methyl-1-octen-6-ynyl]-Δ2(1H),Δ-pentalenevaleric acid, Cilaprost, ZK 36374; (5E)-5-[(3aS,4R,5R,6aS)-Hexahydro-5-hydroxy-4-[(1E,3S)-3-hydroxy-4-methyl-1-octen-6-ynyl]-2(1H)-pentalenylidene]pentanoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H32O4
CAS Number:
Molecular Weight:
360.49
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

film

storage condition

desiccated

color

colorless

solubility

DMSO: 10 mg/mL, clear

storage temp.

−20°C

SMILES string

[H][C@@]12[C@@](C/C(C2)=C/CCCC(O)=O)([H])[C@@H](/C=C/[C@@H](O)C(C)CC#CC)[C@H](O)C1

InChI

1S/C22H32O4/c1-3-4-7-15(2)20(23)11-10-18-19-13-16(8-5-6-9-22(25)26)12-17(19)14-21(18)24/h8,10-11,15,17-21,23-24H,5-7,9,12-14H2,1-2H3,(H,25,26)/b11-10+,16-8+/t15?,17-,18?,19-,20+,21+/m0/s1

InChI key

HIFJCPQKFCZDDL-SGSAMSKHSA-N

Biochem/physiol Actions

Iloprost is a potent prostacyclin PGI2 analog that binds with equal affinity to human prostacyclin (Prostanoid IP) and prostaglandin EP1 receptors. Iloprost acts as a vasodilator and antihypertensive, and inhibits the ADP, thrombin, and collagen-induced aggregation of human platelets. Iloprost dilates systemic and pulmonary arterial vascular beds and has been used clinically for pulmonary arterial hypertension (PAH), scleroderma, and Raynaud′s syndrome.
Iloprost is used to treat children with PH (pulmonary hypertension) after the surgery to clear CHD (congenital heart disease). In transplant patients, this prostacyclin analogue is used to inhibit renal dysfunction. Iloprost performs various functions like vasodilatation and cytoprotection. It has higher stability than prostacyclin.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Iloprost for children with pulmonary hypertension after surgery to correct congenital heart disease.
Xu Z, et al.
Pediatric Pulmonology, 50(6), 588-595 (2015)
Meyler's Side Effects of Endocrine and Metabolic Drugs (2009)
A review of the use of iloprost, a synthetic prostacyclin, in the prevention of radiocontrast nephropathy in patients undergoing coronary angiography and intervention.
Kassis HM, et al.
Clinical Cardiology, 38(8), 492-498 (2015)
Joseph Castillo et al.
Cancer research, 79(15), 3916-3927 (2019-06-12)
Regulatory T cells (Treg) are immunosuppressive and negatively impact response to cancer immunotherapies. CREB-binding protein (CBP) and p300 are closely related acetyltransferases and transcriptional coactivators. Here, we evaluate the mechanisms by which CBP/p300 regulate Treg differentiation and the consequences of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service