Skip to Content
Merck
All Photos(1)

Key Documents

SML1519

Sigma-Aldrich

Macelignan

≥98% (HPLC)

Synonym(s):

(+)-Anwulignan, 4-[(2S,3R)-4-(1,3-Benzodioxol-5-yl)-2,3-dimethylbutyl]-2-methoxy-phenol, Anwulignan

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24O4
CAS Number:
Molecular Weight:
328.40
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

OC1=C(OC)C=C(C[C@H](C)[C@H](C)CC2=CC=C(OCO3)C3=C2)C=C1

InChI

1S/C20H24O4/c1-13(8-15-4-6-17(21)19(10-15)22-3)14(2)9-16-5-7-18-20(11-16)24-12-23-18/h4-7,10-11,13-14,21H,8-9,12H2,1-3H3/t13-,14+/m0/s1

InChI key

QDDILOVMGWUNGD-UONOGXRCSA-N

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
PHL83522A2731SML1766
AMG 9810 ≥98% (HPLC), powder

A2731

AMG 9810

form

powder

form

-

form

powder

form

powder

Quality Level

100

Quality Level

-

Quality Level

100

Quality Level

100

storage temp.

2-8°C

storage temp.

-

storage temp.

2-8°C

storage temp.

2-8°C

solubility

DMSO: 20 mg/mL, clear

solubility

-

solubility

DMSO: >5 mg/mL

solubility

DMSO: 5 mg/mL, clear (warmed)

color

white to beige

color

-

color

white to beige

color

white to light brown

General description

Macelignan is a naturally occurring compound that has scavenging effects on free-radicals and inhibits prostaglandin. It also exhibits antioxidant and anti-inflammatory effects.[1]

Biochem/physiol Actions

Macelignan is a neuroprotective lingan isolated from nutmeg (Myristica fragrans) that protects dopaminergic neurons from inflammatory degeneration. Apparently, the neuroprotective effect of macelignan is mediated by PPARγ activation and arginase-1 expression. Macelignan inhibits LPS induced production of NO as well and inflammatory cytokines including TNF and IL- 1β in primary cultures of microglia.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


  • Certificates of Analysis (COA)

    Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

    Need A Sample COA?

    This is a sample Certificate of Analysis (COA) and may not represent a recently manufactured lot of this specific product.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Chun-Ai Cui et al.
    Neuroscience letters, 448(1), 110-114 (2008-10-23)
    Previous studies have shown that macelignan has anti-inflammatory and neuroprotective effects. Subsequently, in the current study, we demonstrate that oral administrations of macelignan reduce the hippocampal microglial activation induced by chronic infusions of lipopolysaccharide (LPS) into the fourth ventricle of

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service