Skip to Content
Merck
All Photos(1)

Key Documents

SML1382

Sigma-Aldrich

XMD8-92

≥98% (HPLC)

Synonym(s):

2-((2-Ethoxy-4-(4-hydroxypiperidin-1-yl)phenyl)amino)-5,11-dimethyl-5Hbenzo[e]pyrimido[5,4-b][1,4]diazepin-6(11H)-one, 2-[[2-Ethoxy-4-(4-hydroxy-1-piperidinyl)phenyl]amino]-5,11-dihydro-5,11-dimethyl-6H-pyrimido[4,5-b][1,4]benzodiazepin-6-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H30N6O3
CAS Number:
Molecular Weight:
474.55
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 25 mg/mL, clear

storage temp.

2-8°C

SMILES string

OC(CC1)CCN1C2=CC=C(NC3=NC(N(C)C(C=CC=C4)=C4C(N5C)=O)=C5C=N3)C(OCC)=C2

InChI

1S/C26H30N6O3/c1-4-35-23-15-17(32-13-11-18(33)12-14-32)9-10-20(23)28-26-27-16-22-24(29-26)30(2)21-8-6-5-7-19(21)25(34)31(22)3/h5-10,15-16,18,33H,4,11-14H2,1-3H3,(H,27,28,29)

InChI key

QAPAJIZPZGWAND-UHFFFAOYSA-N

Application

XMD8-92 has been used as a specific extracellular signal regulated kinase 5 (ERK5) inhibitor in human umbilical vein endothelial cells (HUVECs) culture.

Biochem/physiol Actions

XMD8-92 is a potent and selective inhibitor of BMK1/ERK5/MAPK7 and DCAMKL1 (DCLK1), kinases implicated in tumorigenesis. XMD8-92 has a Kd of 80 nM for ERK5 and a Kd of 97 nM for DCAMKL1. The next closest targets are DCAMKL2 (190 nM), TNK1 (890 nM), and PLK4 (600 nM). Through inhibition of BMK1 activity, XMD8-92 blocked tumor cell proliferation in vitro in a wide variety of cancer cell lines and significantly inhibited tumor growth in vivo by 95% in mouse studies. XMD8-92 inhibited AsPC-1 human pancreatic cancer cell proliferation and pancreatic tumor xenograft growth via a DCLK1-dependent mechanism.

Other Notes

XMD8-92 has been expertly reviewed and recommended by the Chemical Probes Portal. For more information, please visit the XMD8-92 probe summary on the Chemical Probes Portal website.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

João Paulo M Luiz et al.
Journal of leukocyte biology, 108(4), 1215-1223 (2020-08-04)
Macrophages are highly plastic cells, responding to diverse environmental stimuli to acquire different functional phenotypes. Signaling through MAPKs has been reported to regulate the differentiation of macrophages, but the role of ERK5 in IL-4-mediated M2 macrophage differentiation is still unclear.
Junko Wakao et al.
Biochemical and biophysical research communications, 488(2), 368-373 (2017-05-16)
The skeletal muscle consists of contractile myofibers and plays essential roles for maintenance of body posture, movement, and metabolic regulation. During the development and regeneration of the skeletal muscle tissue, the myoblasts fuse into multinucleated myotubes that subsequently form myofibers.
Hui Wang et al.
International journal of oncology, 51(1), 91-103 (2017-06-01)
Malignant pleural mesothelioma (MPM) is an aggressive cancer for which more effective treatments are needed. In this study, strong to moderate staining of MET and ERK5 was detected in 67.1 and 48% of the analyzed 73 human mesothelioma tumors, and significant
Qiang Li et al.
Journal of clinical neuroscience : official journal of the Neurosurgical Society of Australasia, 34, 264-270 (2016-10-16)
Angiographic vasospasm, especially in the early phases (<72h) of subarachnoid hemorrhage (SAH), is one of the major complications after an aneurysm rupture and is often the cause of delayed neurological deterioration. Scutellarin (SCU), a flavonoid extracted from the traditional Chinese
Joyce K Thompson et al.
Oncotarget, 9(1), 293-305 (2018-02-09)
Malignant mesothelioma is an aggressive cancer in desperate need of treatment. We have previously shown that extracellular signaling regulated kinase 5 (ERK5) plays an important role in mesothelioma pathogenesis using ERK5 silenced human mesothelioma cells exhibiting significantly reduced tumor growth

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service