Skip to Content
Merck
All Photos(1)

Key Documents

SML1327

Sigma-Aldrich

Glutaminase Inhibitor 968

≥98% (HPLC)

Synonym(s):

5-(3-Bromo-4-(dimethylamino)phenyl)-2,2-dimethyl-2,3,5,6-tetrahydrobenzo[a]phenanthridin-4(1H)-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H27BrN2O
CAS Number:
Molecular Weight:
475.42
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

Brc1c(ccc(c1)C2Nc3c(c5c(cc3)cccc5)C4=C2C(=O)CC(C4)(C)C)N(C)C

InChI

1S/C27H27BrN2O/c1-27(2)14-19-24-18-8-6-5-7-16(18)9-11-21(24)29-26(25(19)23(31)15-27)17-10-12-22(30(3)4)20(28)13-17/h5-13,26,29H,14-15H2,1-4H3

InChI key

NVFRRJQWRZFDLM-UHFFFAOYSA-N

General description

Glutaminase Inhibitor 968 belongs to the benzophenanthridinone family.

Application

Glutaminase inhibitor 968 has been used to study the role of glutamine metabolism in macrophage activation.

Biochem/physiol Actions

Glutaminase Inhibitor 968 is an allosteric inihibitor of the mitochondrial enzyme glutaminase C (GAC), which is overexpressed in a number of cancer cell lines. Glutaminase Inhibitor 968 shows 21% inhibition at 10 μM, 94% at 25 μM. Glutaminase Inhibitor 968 blocked human cancer cell proliferation in culture, and in inhibited tumor formation in mouse xenograft models.
Glutaminase Inhibitor 968 is an allosteric inihibitor of the mitochondrial enzyme glutaminase C (GAC).
Variation in the 3-bromo-4-(dimethylamino)phenyl ring of glutaminase inhibitor 968, leads to loss of its inhibitory activity. Glutaminase catalysed glutamine metabolism maintains intracellular pH of cancer cells. Inhibition by glutaminase inhibitor 968 is known to hinder cell proliferation as a consequences of low intracellular pH.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

α-ketoglutarate orchestrates macrophage activation through metabolic and epigenetic reprogramming.
Liu P S, et al.
Nature Immunology, 18(9), 985-985 (2017)
Glutaminase regulation in cancer cells: a druggable chain of events.
Katt W P and Cerione R A
Drug Discovery Today, 19(4), 450-457 (2014)
Glutaminase: a hot spot for regulation of cancer cell metabolism?.
Erickson J W and Cerione R A
Oncotarget, 1(8), 734-734 (2010)
Dibenzophenanthridines as Inhibitors of Glutaminase C and Cancer Cell Proliferation
Katt W P, et al.
Molecular Cancer Therapeutics, 11(6) (2012)
Meijian Liao et al.
EBioMedicine, 41, 200-213 (2019-02-24)
LncRNAs have been found to be involved in various aspects of biological processes. In this study, we aimed to uncover the molecular mechanisms of lncRNA EPB41L4A-AS1 in regulating glycolysis and glutaminolysis in cancer cells. The expression of EPB41L4A-AS1 in cancer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service