Skip to Content
Merck
All Photos(1)

Key Documents

SML1170

Sigma-Aldrich

SR2211

≥98% (HPLC)

Synonym(s):

1,1,1,3,3,3-Hexafluoro-2-(2-fluoro-4-((4-(pyridin-4-ylmethyl)piperazin-1-yl)methyl)-[1,1-biphenyl]-4-yl)propan-2-ol, ;2-Fluoro-4′-[[4-(4-pyridinylmethyl)-1-piperazinyl]methyl]-a,a-bis(trifluoromethyl)-[1,1′-biphenyl]-4-methanol, ML310

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H24F7N3O
CAS Number:
Molecular Weight:
527.48
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

−20°C

SMILES string

FC(F)(F)C(O)(C(F)(F)F)c1cc(c(cc1)c2ccc(cc2)CN3CCN(CC3)Cc4ccncc4)F

InChI

1S/C26H24F7N3O/c27-23-15-21(24(37,25(28,29)30)26(31,32)33)5-6-22(23)20-3-1-18(2-4-20)16-35-11-13-36(14-12-35)17-19-7-9-34-10-8-19/h1-10,15,37H,11-14,16-17H2

InChI key

KVHKWAZUPPBMLL-UHFFFAOYSA-N

Biochem/physiol Actions

SR2211 (ML310) is a potent and selective inverse agonist of retinoic acid receptor-related orphan receptor gamma (RORγ). RORγ is essential for interleukin 17 expression and differentiation of TH17 cells, important in autoimmune diseases including multiple sclerosis, rheumatoid arthritis, inflammatory bowel disease and lupus. SR2211 (ML310) treatment of EL-4 murine T lymphocyte cells resulted in suppression of gene expression and production of IL-17 with an IC50 ~320 nM with no effect on the transcriptional activity of RORγ. In a mouse model of rheumatoid arthritis, SR2211 significantly reduced joint inflammation, suppressing inflammatory T cell function, expression and production of inflammatory cytokines, and macrophage activation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service