Skip to Content
Merck
All Photos(1)

Documents

SML1141

Sigma-Aldrich

Solasodine

≥95% (HPLC)

Synonym(s):

(-)-Solasodine, (3β,22α,25R)-Spirosol-5-en-3-ol, NSC 178260, NSC 179187, Purapuridine, Solancarpidine, Solasod-5-en-3β-ol, Solasod-5-en-3β-ol (7CI,8CI), Solasodin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H43NO2
CAS Number:
Molecular Weight:
413.64
EC Number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

optical activity

[α]/D -88 to -98°, c = 0.2 in methanol

color

white to beige

solubility

DMSO: 0.5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

C[C@@H]1[C@]2(NC[C@H](C)CC2)O[C@@]3([H])C[C@@]4([H])[C@]5([H])CC=C6C[C@@H](O)CC[C@]6(C)[C@@]5([H])CC[C@]4(C)[C@]31[H]

InChI

1S/C27H43NO2/c1-16-7-12-27(28-15-16)17(2)24-23(30-27)14-22-20-6-5-18-13-19(29)8-10-25(18,3)21(20)9-11-26(22,24)4/h5,16-17,19-24,28-29H,6-15H2,1-4H3/t16-,17+,19+,20-,21+,22+,23+,24+,25+,26+,27-/m1/s1

InChI key

KWVISVAMQJWJSZ-VKROHFNGSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Solasodine has been used in viability assays.

Biochem/physiol Actions

Solasodine is a neuroprotective antioxidant glycoalkaloid of Solanum species. Solasodine increases superoxide dismutase (SOD), catalase (CAT),and glutathione (GSH) levels, while reducing lipid peroxidation (LPO) and nitric oxide (NO) levels in the brains of ischemia/reperfusion (I/R)-injury model in rats. Also, Solasonine displayed multiple activities including, anti-cancer and leishmanicidal activities.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Agave negatively regulates YAP and TAZ transcriptionally and post-translationally in osteosarcoma cell lines
Ferraiuolo M, et al.
Cancer letters (2018)
Agave negatively regulates YAP and TAZ transcriptionally and post-translationally in osteosarcoma cell lines
Ferraiuolo M, et al.
Cancer Letters (2018)

Articles

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

DISCOVER Bioactive Small Molecules for Nitric Oxide & Cell Stress Research

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service