Skip to Content
Merck
All Photos(1)

Key Documents

SML1043

Sigma-Aldrich

KC7F2

≥98% (HPLC)

Synonym(s):

N,N′-(Dithiodi-2,1-ethanediyl)bis[2,5-dichloro-benzenesulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H16Cl4N2O4S4
CAS Number:
Molecular Weight:
570.38
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

shipped in

wet ice

storage temp.

−20°C

SMILES string

ClC1=CC=C(Cl)C=C1S(NCCSSCCNS(C2=C(Cl)C=CC(Cl)=C2)(=O)=O)(=O)=O

InChI

1S/C16H16Cl4N2O4S4/c17-11-1-3-13(19)15(9-11)29(23,24)21-5-7-27-28-8-6-22-30(25,26)16-10-12(18)2-4-14(16)20/h1-4,9-10,21-22H,5-8H2

InChI key

REQLACDIZMLXIC-UHFFFAOYSA-N

Biochem/physiol Actions

KC7F2 is a cell permeable potent HIF-1 pathway inhibitor.
KC7F2 is a cell permeable potent HIF-1 pathway inhibitor. KC7F2 inhibits the activation of HIF-target genes such as carbonic anhydrase IX, matrix metalloproteinase 2 (MMP2), enolase 1, and endothelin 1. KC7F2 suppresses the protein accumulation of HIF-1? in cancer cell lines by inhibition of its protein synthesis at the translation level. KC7F2 is preferentially cytotoxicity to cancer cells, an effect that is increased in hypoxia.

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Other Notes

air sensitive

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yao Wang et al.
Acta biochimica et biophysica Sinica, 50(3), 281-287 (2018-01-03)
High-glucose level exerts deleterious effects on pancreatic β cells, but the mechanisms remain unclear. Calcium/calmodulin-dependent serine protein kinase (CASK) plays a vital role in neural development and release of neurotransmitters, and probably plays a role in the anchoring of insulin
Jiao Cheng et al.
International journal of molecular medicine, 43(2), 945-955 (2018-12-21)
Angiogenesis is an essential process involved in various physiological, including placentation, and pathological, including cancer and endometriosis, processes. Melatonin (MLT), a well‑known natural hormone secreted primarily in the pineal gland, is involved in regulating neoangiogenesis and inhibiting the development of
Chengjun Yuan et al.
Die Pharmazie, 74(3), 157-162 (2019-04-10)
The aim of this study was to observe the effects of HIF-1α activation on myocardial I/R in diabetes. Diabetes was induced in an experimental rat model, and regulators of HIF-1α including KC7F2, deferoxamine and ginsenoside Rg1 were administered to observe
Yun-Sung Lim et al.
Anticancer research, 37(2), 599-606 (2017-02-10)
Tumor hypoxia induces hypoxia-inducible factor-1α (HIF1α), which can influence tumorigenesis and metastasis. We evaluated the expression of HIF1α and the effect of HIF1α inhibitors in adenoid cystic carcinoma (ACC). HIF1α expression was demonstrated in ACC cell lines (ACC2 and ACCM).
Guofang Chen et al.
Oncogenesis, 9(9), 81-81 (2020-09-12)
Endometrial cancer stem cells (ECSCs) are stem-like cells endowed with self-renewal and differentiation abilities, and these cells are essential for cancer progression in endometrial cancer (EC). As hallmarks of the tumour microenvironment (TME), hypoxia and hypoxia-inducing factors (HIFs) give rise

Articles

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service