Skip to Content
Merck
All Photos(1)

Key Documents

SML1023

Sigma-Aldrich

Flucloxacillin sodium

≥95% (HPLC)

Synonym(s):

(2S,5R,6R)-6-[[[3-(2-Chloro-6-fluorophenyl)-5-methyl-4-isoxazolyl]carbonyl]amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid sodium, BRL 2039 sodium, FK 900 sodium, Floxacillin sodium

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H16ClFN3O5S · Na
CAS Number:
Molecular Weight:
475.85
EC Number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

storage condition

protect from light

color

white to beige

solubility

H2O: 20 mg/mL, clear

storage temp.

−20°C

SMILES string

[Na+].Fc1c(c(ccc1)Cl)c2n[o]c(c2\C(=N\[C@H]3[C@H]4SC([C@@H](N4C3=O)C(=O)O)(C)C)\[O-])C

InChI

1S/C19H17ClFN3O5S.Na/c1-7-10(12(23-29-7)11-8(20)5-4-6-9(11)21)15(25)22-13-16(26)24-14(18(27)28)19(2,3)30-17(13)24;/h4-6,13-14,17H,1-3H3,(H,22,25)(H,27,28);/q;+1/p-1/t13-,14+,17-;/m1./s1

InChI key

OTEANHMVDHZOPB-SLINCCQESA-M

General description

Flucloxacillin sodium, a sodium salt of flucloxacillin, is a member of the penicillin class.

Application

Flucloxacillin sodium has been used as a β-lactam antibiotic to study its effects on methicillin-resistant Staphylococcus aureus (MRSA) ΔfloA mutant.

Biochem/physiol Actions

Flucloxacillin exhibits antimicrobial property.
Flucloxacillin is a narrow-spectrum β-lactam antibiotic used to treat Gram-positive bacterial infections. Flucloxacillin induces cholestatic liver damage.
Flucloxacillin is a narrow-spectrum β-lactam antibiotic.

Other Notes

Light sensitive

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Handbook of Pharmaceutical Excipients (2014)
Apparent Molar Volume, Adiabatic Compressibility, and Critical Micelle Concentration of Flucloxacillin Sodium in Aqueous NaCl Solutions at Different Temperatures
Khatun MR, et al.
Journal of Chemical and Engineering Data, 61(1), 102-113 (2016)
Membrane microdomain disassembly inhibits MRSA antibiotic resistance
Garcaa-Fernandez E, et al.
Cell, 171(6), 1354-1367 (2017)
Esther García-Fernández et al.
Cell, 171(6), 1354-1367 (2017-11-07)
A number of bacterial cell processes are confined functional membrane microdomains (FMMs), structurally and functionally similar to lipid rafts of eukaryotic cells. How bacteria organize these intricate platforms and what their biological significance is remain important questions. Using the pathogen
Ryan Nattrass et al.
Toxicological sciences : an official journal of the Society of Toxicology, 146(1), 146-156 (2015-04-17)
There are currently no animal models of drug-induced liver injury (DILI) where the adaptive immune system has been shown to damage the liver. Thus, it is difficult to explore the mechanistic basis of the tissue injury. The aim of this

Articles

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Bioactive small molecules for immune system signaling target identification/validation and antibiotics, antivirals, and antifungals offered.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service