Skip to Content
Merck

SML0917

BMS-986122

≥98% (HPLC)

Synonym(s):

2-(3-Bromo-4-methoxyphenyl)-3-[(4-chlorophenyl)sulfonyl]-thiazolidine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C16H15BrClNO3S2
CAS Number:
Molecular Weight:
448.78
NACRES:
NA.77
UNSPSC Code:
12352200
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

BMS-986122 is a positive allosteric modulator (PAM of the m-opioid receptor). BMS986122 displays little or no agonist activity alone, but dose dependently increases endomorphin-I induced b-arrestin recruitment, and inhibition of forskolin-induced adenyl cyclase activity. The compound also potentiates DAMGO-stimulated GTPgS receptor binding.
BMS-986122 is a positive allosteric modulator (PAM of the m-opioid receptor.
BMS-986122 may exhibit an antinociception effect in vivo by increasing the efficiency of Met-enkephalin (met-Enk) to inhibit γ aminobutyric acid (GABA) release in the periaqueductal gray region of the brain.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.


Hazard Codes

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Eye Irrit. 2

Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library