Skip to Content
Merck
All Photos(1)

Key Documents

SML0797

Sigma-Aldrich

Flibanserin

≥98% (HPLC)

Synonym(s):

1,3-Dihydro-1-[2-[4-[3-(trifluoromethyl)phenyl]-1-piperazinyl]ethyl]-2H-benzimidazol-2-one, BIMT 17, BIMT 17BS

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H21F3N4O
CAS Number:
Molecular Weight:
390.40
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

FC(F)(F)c1ccc(cc1)N2CCN(CC2)CC[n]3c4c([nH][c]3=O)cccc4

InChI

1S/C20H21F3N4O/c21-20(22,23)15-5-7-16(8-6-15)26-12-9-25(10-13-26)11-14-27-18-4-2-1-3-17(18)24-19(27)28/h1-8H,9-14H2,(H,24,28)

InChI key

CCIZQGRSEAHATK-UHFFFAOYSA-N

Biochem/physiol Actions

Flibanserin is a 5-HT1A receptor agonist and 5-HT2A receptor antagonist.
Flibanserin is a 5-HT1A receptor agonist and 5-HT2A receptor antagonist. Flibanserin has high affinity for human 5-HT1A receptors (Ki = 1 nm) and lower affinity for 5-HT2A (Ki = 49 nm) and D4 (Ki = 4–24 nm) receptors, and negligible affinity for a variety of other neurotransmitter receptors and ion channels. Flibanserin was investigated as a novel, non-hormonal treatment for pre-menopausal women with Hypoactive Sexual Desire Disorder (HSDD). Flibanserin. Recently, Flibanserin was found to reduce L-DOPA-induced dyskinesia in a model of Parkinson′s Disease.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sidney Kennedy
The journal of sexual medicine, 7(10), 3449-3459 (2010-07-22)
Flibanserin, a novel 5-HT(1A) agonist and 5-HT(2A) antagonist, has the potential to treat sexual dysfunction. Provide historical perspective on the rationale for development of flibanserin to treat sexual dysfunction, based on post hoc analyses of data. The Arizona Sexual Experiences
Boehringer Ingelheim withdraws libido drug for women.
Jeanne Lenzer
BMJ (Clinical research ed.), 341, c5701-c5701 (2010-10-14)
John Thorp et al.
The journal of sexual medicine, 9(3), 793-804 (2012-01-14)
Hypoactive Sexual Desire Disorder (HSDD) is characterized by low sexual desire that causes marked distress or interpersonal difficulty. To assess the efficacy and tolerability of flibanserin, a postsynaptic 5-HT1A agonist/5-HT2A antagonist, in the treatment of premenopausal women with HSDD. North
Drug for low sexual desire carries significant harms, FDA advisers find.
Ray Moynihan
BMJ (Clinical research ed.), 341, c3339-c3339 (2010-07-09)
Dysfunctional women remain unsexed.
Michael G Wyllie
BJU international, 106(3), 431-432 (2010-07-27)

Articles

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service