Skip to Content
Merck
All Photos(1)

Key Documents

SML0770

Sigma-Aldrich

P5091

≥98% (HPLC)

Synonym(s):

1-[5-[(2,3-Dichlorophenyl)thio]-4-nitro-2-thienyl]-ethanone; 1-[5-(2,3-dichlorophenyl)sulfanyl-4-nitro-2-thienyl]ethanone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H7Cl2NO3S2
CAS Number:
Molecular Weight:
348.22
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

light yellow to dark yellow

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

[s]1c(c(cc1C(=O)C)[N+](=O)[O-])Sc2c(c(ccc2)Cl)Cl

InChI

1S/C12H7Cl2NO3S2/c1-6(16)10-5-8(15(17)18)12(20-10)19-9-4-2-3-7(13)11(9)14/h2-5H,1H3

InChI key

LKZLGMAAKNEGCH-UHFFFAOYSA-N

Application

P5091 has been used:
  • as a ubiquitin specific peptidase 47 (USP47) inhibitor in 3-(4,5-dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT) assay to evaluate the cellular viability of MCF-10A cells
  • as a USP7 inhibitor to study the regulatory role for USP7 on inflammasome activation
  • as USP7 inhibitor in drug susceptibility assays to study its effect on bone marrow−resident tumor cells (BMRTCs)/ circulating tumor cells (CTCs)

Biochem/physiol Actions

P5091 is a cell permeable, potent and specific inhibitor of deubiquitylating enzyme USP7 (Ubiquitin-Specific Protease-7) that induces HDM2 polyubiquitylation and accelerates degradation of HDM2. P5091 induces apoptosis in MM cells resistant to conventional and bortezomib therapies. P5091 inhibits tumor growth and prolongs survival in animal models of cancer.
P5091 is a cell permeable, potent and specific inhibitor of deubiquitylating enzyme USP7.
P5091 plays an important role in ovarian cancer, as it can prevent the growth of cells and can promote necrosis and apoptosis.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 4

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

11th Int. Conf. Adv. Prostagl. Leukot. Res. null
13th ACS Winter Fluorine Conference, paper No. 8 null
12th International Detonation Symposium. null
13th ACS Winter Fluorine Conference, paper No. 8 null
Sandhini Saha et al.
Cell death & disease, 13(6), 563-563 (2022-06-23)
Nutrient surplus and consequent free fatty acid accumulation in the liver cause hepatosteatosis. The exposure of free fatty acids to cultured hepatocyte and hepatocellular carcinoma cell lines induces cellular stress, organelle adaptation, and subsequent cell death. Despite many studies, the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service