Skip to Content
Merck
All Photos(1)

Key Documents

SML0667

Sigma-Aldrich

PF-429242 dihydrochloride

≥98% (HPLC)

Synonym(s):

4-[(Diethylamino)methyl]-N-[2-(2-methoxyphenyl)ethyl]-N-(3R)-3-pyrrolidinyl-benzamide dihydrochloride, PF 429242 dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H35N3O2 · 2HCl
CAS Number:
Molecular Weight:
482.49
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 15 mg/mL, clear

storage temp.

2-8°C

SMILES string

N1C[C@@H](CC1)N(CCc3c(cccc3)OC)C(=O)c2ccc(cc2)CN(CC)CC

InChI

1S/C25H35N3O2/c1-4-27(5-2)19-20-10-12-22(13-11-20)25(29)28(23-14-16-26-18-23)17-15-21-8-6-7-9-24(21)30-3/h6-13,23,26H,4-5,14-19H2,1-3H3/t23-/m1/s1

InChI key

XKPJTOHUPQWSOJ-HSZRJFAPSA-N

Application

PF-429242 dihydrochloride has been used as an inhibitor of sterol regulatory element-binding protein (SREBP).

Biochem/physiol Actions

PF-429242 is a potent inhibitor of S1P (cellular proprotein convertase sterol regulatory element-binding protein (SREBP) site 1 protease) that reduces expression levels of hepatic SREBP target genes and lower rates of cholesterol and fatty acid synthesis in mice. PF-429242 is a potent antiviral agent against prototypic arenavirus lymphocytic choriomeningitis virus (LCMV) and LASV in cultured cells. PF-429242 efficiently prevented the processing of GPC from the LCMV and LASV. PF-429242 is expected to be active against CCHFV.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Siqingaowa et al.
Biochemical and biophysical research communications, 488(1), 136-140 (2017-05-10)
Sterol regulatory element-binding protein1 (SREBP1) is a key regulatory factor that controls lipid homeostasis. Overactivation of SREBP1 and elevated lipid biogenesis are considered the major characteristics in malignancies of prostate cancer, endometrial cancer, and glioblastoma. However, the impact of SREBP1
Ruofan Wang et al.
Cell, 184(1), 106-119 (2020-12-18)
The Coronaviridae are a family of viruses that cause disease in humans ranging from mild respiratory infection to potentially lethal acute respiratory distress syndrome. Finding host factors common to multiple coronaviruses could facilitate the development of therapies to combat current
Fatostatin inhibits cancer cell proliferation by affecting mitotic microtubule spindle assembly and cell division.
Gholkar A A, et al.
The Journal of Biological Chemistry, 291(33), 17001-17008 (2016)
Tian Du et al.
Breast cancer research : BCR, 20(1), 106-106 (2018-09-06)
Invasive lobular breast carcinoma (ILC) is a histological subtype of breast cancer that is characterized by loss of E-cadherin and high expression of estrogen receptor alpha (ERα). In many cases, ILC is effectively treated with adjuvant aromatase inhibitors (AIs); however
Jessica D Rosarda et al.
ACS chemical biology, 16(12), 2852-2863 (2021-11-20)
The extracellular accumulation of glutamate is a pathologic hallmark of numerous neurodegenerative diseases including ischemic stroke and Alzheimer's disease. At high extracellular concentrations, glutamate causes neuronal damage by promoting oxidative stress, which can lead to cellular death. This has led

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service