Skip to Content
Merck
All Photos(1)

Key Documents

SML0620

Sigma-Aldrich

AF-DX 384 Free Base

≥98% (HPLC)

Synonym(s):

N-[2-[2-[(Dipropylamino)methyl]-1-piperidinyl]ethyl]-5,6-dihydro-6-oxo-11H-pyrido[2,3-b][1,4]benzodiazepine-11-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H38N6O2
CAS Number:
Molecular Weight:
478.63
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear (warmed)

storage temp.

room temp

SMILES string

N4(C(CCCC4)CN(CCC)CCC)CCNC(=O)[n]1c2ncccc2[nH][c](c3c1cccc3)=O

InChI

1S/C27H38N6O2/c1-3-16-31(17-4-2)20-21-10-7-8-18-32(21)19-15-29-27(35)33-24-13-6-5-11-22(24)26(34)30-23-12-9-14-28-25(23)33/h5-6,9,11-14,21H,3-4,7-8,10,15-20H2,1-2H3,(H,29,35)(H,30,34)

InChI key

MZDYABXXPZNUCT-UHFFFAOYSA-N

Biochem/physiol Actions

AF-DX 384 Free Base is a potent muscarinic M2/M4 receptor selective antagonist (pKi values are 8.22, 8.00, 7.51, 7.18 and 6.27 at human M2, M4, M1, M3 and M5 receptors respectively).

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the Acetylcholine Receptors (Muscarinic) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service