Skip to Content
Merck
All Photos(1)

Key Documents

SML0616

Sigma-Aldrich

nTZDpa

≥98% (HPLC)

Synonym(s):

5-Chloro-1-[(4-chlorophenyl)methyl]-3-(phenylthio)-1H-indole-2-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H15Cl2NO2S
CAS Number:
Molecular Weight:
428.33
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 15 mg/mL, clear

storage temp.

2-8°C

InChI

1S/C22H15Cl2NO2S/c23-15-8-6-14(7-9-15)13-25-19-11-10-16(24)12-18(19)21(20(25)22(26)27)28-17-4-2-1-3-5-17/h1-12H,13H2,(H,26,27)

InChI key

VUPOTURDKDMIGQ-UHFFFAOYSA-N

Biochem/physiol Actions

nTZDpa is a non-thiazolidinedione PPARγ partial agonist and selective PPARγ modulator (SPPARγM). nTZDpa binds to PPARγ with high affinity, but partially activates the receptor in cell-based assays. nTZDpa reduces insulin resistance and hyperglycemia in obese diabetic rodents equal to TZD full agonists, but without increasing cardiac weight and adiposity.

Features and Benefits

This compound is featured on the Nuclear Receptors (Non-Steroids) and Nuclear Receptors (PPARs) pages of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hang Yuan et al.
International journal of molecular sciences, 22(16) (2021-08-28)
The periodontal ligament is a soft connective tissue embedded between the alveolar bone and cementum, the surface hard tissue of teeth. Periodontal ligament fibroblasts (PDLF) actively express osteo/cementogenic genes, which contribute to periodontal tissue homeostasis. However, the key factors maintaining

Articles

Peroxisome proliferator activated receptors (PPARs) are ligand-activated transcription factors related to hormone receptors, influencing gene expression.

Peroxisome proliferator activated receptors (PPARs) are ligand-activated transcription factors related to hormone receptors, influencing gene expression.

Peroxisome proliferator activated receptors (PPARs) are ligand-activated transcription factors related to hormone receptors, influencing gene expression.

Peroxisome proliferator activated receptors (PPARs) are ligand-activated transcription factors related to hormone receptors, influencing gene expression.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service