Skip to Content
Merck
All Photos(1)

Key Documents

SML0594

Sigma-Aldrich

CGP 55845 hydrochloride

≥98% (HPLC)

Synonym(s):

(2S)-3-[[(1S)-1-(3,4-Dichlorophenyl)ethyl]amino-2-hydro xypropyl](phenylmethyl)phosphinic acid hydrochloride, CGP 55845A, CGP55845

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H22Cl2NO3P · HCl
CAS Number:
Molecular Weight:
438.71
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

room temp

SMILES string

[P](=O)(O)(C[C@@H](O)CN[C@@H](C)c2cc(c(cc2)Cl)Cl)Cc1ccccc1

InChI

1S/C18H22Cl2NO3P/c1-13(15-7-8-17(19)18(20)9-15)21-10-16(22)12-25(23,24)11-14-5-3-2-4-6-14/h2-9,13,16,21-22H,10-12H2,1H3,(H,23,24)/t13-,16-/m0/s1

InChI key

ZODSPDOOCZZEIM-BBRMVZONSA-N

Biochem/physiol Actions

CGP 55845 is a potent selective GABA-B receptor antagonist with an IC50 of 5 nM.
CGP 55845 is a potent selective GABA-B receptor antagonist..

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the GABAB Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ahmad A Khundakar et al.
European journal of pharmacology, 671(1-3), 33-38 (2011-09-21)
Brain-derived neurotrophic factor (BDNF) has been suggested as a target for antidepressant treatment and chronic antidepressant drug administration shows a 'biphasic effect' on BDNF mRNA in rat hippocampus (transient decrease followed by an increase). In comparison, following acute administration only
Wendy W Wu et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 31(7), 2638-2648 (2011-02-18)
Premature and uncompensated loss of ovarian hormones following ovariectomy (OVX) elevates the risks of cognitive impairment and dementia. These risks are prevented with estrogen (E(2))-containing hormone replacement therapy initiated shortly following OVX but not after substantial delay. Currently, the cellular
Mark O Cunningham et al.
Epilepsia, 53(7), 1205-1214 (2012-06-13)
We describe a form of very fast oscillation (VFO) in patient electrocorticography (ECoG) recordings, that can occur prior to ictal events, in which the frequency increases steadily from ≈ 30-40 to >120 Hz, over a period of seconds. We dub
Dustin Anderson et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 33(18), 7811-7824 (2013-05-03)
Synaptic transmission and neuronal excitability depend on the concentration of extracellular calcium ([Ca](o)), yet repetitive synaptic input is known to decrease [Ca](o) in numerous brain regions. In the cerebellar molecular layer, synaptic input reduces [Ca](o) by up to 0.4 mm
Volodymyr Dzhala et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 32(12), 4017-4031 (2012-03-24)
Severe head trauma causes widespread neuronal shear injuries and acute seizures. Shearing of neural processes might contribute to seizures by disrupting the transmembrane ion gradients that subserve normal synaptic signaling. To test this possibility, we investigated changes in intracellular chloride

Articles

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service