Skip to Content
Merck
All Photos(1)

Documents

SML0560

Sigma-Aldrich

BRACO19 hydrochloride

≥96% (HPLC), powder, telomerase inhibitor

Synonym(s):

N,N′-(9-(4-(Dimethylamino)phenylamino)acridine-3,6-diyl)bis(3-(pyrrolidin-1-yl)propanamide) hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H43N7O2 · xHCl
CAS Number:
Molecular Weight:
593.76 (free base basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

product name

BRACO19 hydrochloride, ≥96% (HPLC)

Quality Level

Assay

≥96% (HPLC)

form

powder

storage condition

desiccated

color

yellow to brown

solubility

H2O: 5 mg/mL, clear

storage temp.

2-8°C

SMILES string

Cl.Cl.Cl.CN(C)c1ccc(Nc2c3ccc(NC(=O)CCN4CCCC4)cc3nc5cc(NC(=O)CCN6CCCC6)ccc25)cc1

InChI

1S/C35H43N7O2.3ClH/c1-40(2)28-11-7-25(8-12-28)38-35-29-13-9-26(36-33(43)15-21-41-17-3-4-18-41)23-31(29)39-32-24-27(10-14-30(32)35)37-34(44)16-22-42-19-5-6-20-42;;;/h7-14,23-24H,3-6,15-22H2,1-2H3,(H,36,43)(H,37,44)(H,38,39);3*1H

InChI key

MJAPNWJRLLDPAB-UHFFFAOYSA-N

Application

BRACO19 hydrochloride has been used to study the role of antiviral activity of G4-stabilizing agents in cells infected with latent human immunodeficiency virus -1 (HIV-1).

Biochem/physiol Actions

BRACO19 is a telomerase inhibitor that stabilizes G-quadruplexes, targeting telomeric G-quadruplexes, inducing DNA damage and cell-cycle arrest.
Telomerase is associated with tumor progression and is highly expressed in a number of tumors. BRACO19 is known to prevent the capping and catalytic action of telomerase and thus, might serve as an effective therapeutic approach for cancer treatment. BRACO19 establishes antiviral activity against HIV (human immunodeficiency virus).

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anti-HIV-1 activity of the G-quadruplex ligand BRACO-19.
Perrone R, et al.
The Journal of Antimicrobial Chemotherapy, 69(12), 3248-3258 (2014)
The G-quadruplex-interactive molecule BRACO-19 inhibits tumor growth, consistent with telomere targeting and interference with telomerase function.
Burger A M, et al.
Cancer Research, 65(4), 1489-1496 (2005)
Yu-Lun Chen et al.
The Chinese journal of physiology, 59(6), 331-347 (2016-11-07)
Lung resistance-related protein (LRP) is a human major vault protein (MVP) implicated in drug resistance of cancer cells in a cell-type dependent manner. The primary goal of the study was to understand the role(s) of LRP in doxorubicin (DOX) resistance
Deficiency in DNA damage response, a new characteristic of cells infected with latent HIV-1.
Piekna-Przybylska D, et al.
Cell Cycle, 16(10), 968-978 (2017)
Shyam Sushama Jose et al.
Frontiers in genetics, 9, 345-345 (2018-09-14)
Telomeropathies are rare disorders associated with impaired telomere length control mechanisms that frequently result from genetic mutations in the telomerase complex. Dyskeratosis congenita is a congenital progressive telomeropathy in which mutation in the telomerase RNA component (TERC) impairs telomere maintenance

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service