Skip to Content
Merck
All Photos(1)

Key Documents

SML0043

Sigma-Aldrich

Racecadotril

≥98% (HPLC)

Synonym(s):

(RS)-Benzyl N-[3-(acetylthio)-2-benzylpropanoyl]glycinate, (±)-Acetorphan, Acetorphan, Cadotril, Redotil, Tiorfan, Zedott

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H23NO4S
CAS Number:
Molecular Weight:
385.48
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to tan

solubility

DMSO: ≥44 mg/mL

storage temp.

room temp

SMILES string

CC(=O)SCC(Cc1ccccc1)C(=O)NCC(=O)OCc2ccccc2

InChI

1S/C21H23NO4S/c1-16(23)27-15-19(12-17-8-4-2-5-9-17)21(25)22-13-20(24)26-14-18-10-6-3-7-11-18/h2-11,19H,12-15H2,1H3,(H,22,25)

InChI key

ODUOJXZPIYUATO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Racecadotril may be used to study enkephalinase-mediated cellular changes.

Biochem/physiol Actions

Inhibition of enkephalinase by Racecadotril prevents the breakdown of enkephalins in the gastrointestinal tract. This reduces secretion of water and electrolytes into the gut without affecting the mobility.
Neutral endopeptidase inhibitor; antidiarrheal
Racecadotril is a neutral endopeptidase inhibitor, an antidiarrheal drug which acts as a peripherally acting enkephalinase inhibitor.

Features and Benefits

This compound is featured on the Neuropeptidases page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hypersensitivity to racecadotril: a case report.
Eleonora Nucera et al.
European journal of pediatrics, 165(6), 418-419 (2006-05-02)
Alfredo Guarino et al.
Journal of pediatric gastroenterology and nutrition, 48 Suppl 2, S58-S62 (2009-03-28)
The colon is actively implicated in intestinal infections not only as a target of enteric pathogens and their products but also as a target organ for treatment. In the presence of diarrhea, both of osmotic and secretory nature, the colon
K Mallikarjun Reddy et al.
Die Pharmazie, 61(12), 994-998 (2007-02-08)
Three unknown impurities in racecadotril bulk drug at levels below 0.5% were detected by simple reverse phase isocratic high performance liquid chromatography (HPLC). Structures for these impurities were proposed by molecular ion information and their fragmentation pattern obtained by LC-MS
Reshma S Baliga et al.
American journal of respiratory and critical care medicine, 178(8), 861-869 (2008-08-12)
Phosphodiesterase 5 (PDE5) inhibitors (e.g., sildenafil) are selective pulmonary vasodilators in patients with pulmonary arterial hypertension. The mechanism(s) underlying this specificity remains unclear, but studies in genetically modified animals suggest it might be dependent on natriuretic peptide bioactivity. We explored
Mar Santos et al.
The Journal of pediatrics, 155(1), 62-67 (2009-04-28)
To compare the efficacy of therapy with racecadotril plus oral rehydration versus oral rehydration alone in children with gastroenteritis in an outpatient setting care. Prospective, randomized, open and parallel study performed in a Pediatric Emergency Service of a tertiary care

Articles

We offers many products related to neuropeptidases for your research needs.

We offers many products related to neuropeptidases for your research needs.

We offers many products related to neuropeptidases for your research needs.

We offers many products related to neuropeptidases for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service