Skip to Content
Merck
All Photos(1)

Key Documents

SML0036

Sigma-Aldrich

Procarbazine hydrochloride

≥98% (HPLC)

Synonym(s):

N-(1-Methylethyl)-4-[(2-methylhydrazinyl)methyl]benzamide hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H19N3O·HCl
CAS Number:
Molecular Weight:
257.76
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to tan

solubility

DMSO: ≥18 mg/mL

storage temp.

2-8°C

SMILES string

Cl.CNNCc1ccc(cc1)C(=O)NC(C)C

InChI

1S/C12H19N3O.ClH/c1-9(2)15-12(16)11-6-4-10(5-7-11)8-14-13-3;/h4-7,9,13-14H,8H2,1-3H3,(H,15,16);1H

InChI key

DERJYEZSLHIUKF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Procarbazine hydrochloride has been used to study its physicochemical property.
Procarbazine, an antineoplastic alkylating agent, may be used to study its pharmacokinetics, metabolism, safety, efficacy and methods of delivery as an anticancer agent. It may be used to study new combination of anticancer drugs.

Biochem/physiol Actions

Procarbazine hydrochloride is a Antineoplastic alkylating agent
Procarbazine is an antineoplastic alkylating agent widely used in cancer chemotherapy in combination with other compounds. It has multiple mechanisms of action. Procarbazine inhibits protein, RNA and DNA synthesis in addition to being an alkylating agent.
Procarbazine is metabolized to azoprocarbazine either by cytochrome P450 enzyme and monoamine oxidase in an NADPH (nicotinamide adenine diphosphate) dependent or independent manner. It is considered neurotoxic. Procarbazine is used in combination with therapy for treating Hodgkin′s disease.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 1B - Muta. 2 - Repr. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Fourteenth biannual report of the Cochrane Haematological Malignancies Group--focus on autologous stem cell transplantation in hematological malignancies.
Michaela Rancea et al.
Journal of the National Cancer Institute, 104(14), NP-NP (2012-07-25)
Anti-Cancer Drug Design, 143-143 (1994)
Mary R Welch et al.
Neuro-oncology, 14(10), 1304-1311 (2012-09-07)
Up to 20% of all primary CNS lymphoma (PCNLS) patients are aged 80 years or older, yet data are limited on how best to treat this rapidly growing population. Despite demographic pressures and the proven efficacy of methotrexate (MTX)-based regimens
L Kochbati et al.
Cancer radiotherapie : journal de la Societe francaise de radiotherapie oncologique, 16(7), 627-632 (2012-10-23)
To assess the epidemiological, clinical and therapeutic aspects of pediatric Hodgkin disease (HD) and to study prognosis factors. We retrospectively reviewed the medical records of children (≤18 years) with HD treated between 1st January 1994 and 31st December 2004. Chemotherapy
Lale Erdem-Eraslan et al.
Journal of clinical oncology : official journal of the American Society of Clinical Oncology, 31(3), 328-336 (2012-12-28)
Intrinsic glioma subtypes (IGSs) are molecularly similar tumors that can be identified based on unsupervised gene expression analysis. Here, we have evaluated the clinical relevance of these subtypes within European Organisation for Research and Treatment of Cancer (EORTC) 26951, a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service