Skip to Content
Merck
All Photos(1)

Key Documents

SMB00119

Sigma-Aldrich

Arjunolic acid

≥95% (LC/MS-ELSD)

Synonym(s):

2,3,23-Trihydroxyolean-12-en-28-oic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H48O5
CAS Number:
Molecular Weight:
488.70
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥95% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

CC1(C)CC[C@@]2(CC[C@]3(C)C(=CC[C@@H]4[C@@]5(C)C[C@@H](O)[C@H](O)[C@@](C)(CO)[C@@H]5CC[C@@]34C)[C@@H]2C1)C(O)=O

InChI

1S/C30H48O5/c1-25(2)11-13-30(24(34)35)14-12-28(5)18(19(30)15-25)7-8-22-26(3)16-20(32)23(33)27(4,17-31)21(26)9-10-29(22,28)6/h7,19-23,31-33H,8-17H2,1-6H3,(H,34,35)/t19-,20+,21+,22+,23-,26-,27-,28+,29+,30-/m0/s1

InChI key

RWNHLTKFBKYDOJ-DDHMHSPCSA-N

General description

Natural product derived from plant source.

Biochem/physiol Actions

Arjunolic acid (AA) is a triterpenoid saponin that has known anti-oxidant activity. AA has also shown promise in against type 1 diabetes through its prevention of hyperglycemia induced activation of MAPKs, PKC, NF-kB signaling cascades. Arjunolic acid is beneficial in protecting again acetaminophen (APAP)-induced liver and hepatocyte injury.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jyotirmoy Ghosh et al.
Free radical biology & medicine, 48(4), 535-553 (2009-12-09)
Acetaminophen (APAP) is a widely used analgesic and antipyretic drug and is safe at therapeutic doses but its overdose frequently causes liver injury. In earlier studies, we demonstrated that arjunolic acid (AA) has a protective effect against chemically induced hepatotoxicity.
Prasenjit Manna et al.
Toxicology, 257(1-2), 53-63 (2009-01-10)
Increasing evidences in both experimental and clinical studies suggest that oxidative stress is involved in the pathogenesis of diabetic tissue damage. Pancreatic beta-cell death is the cause of decreased insulin production in diabetes. Streptozotocin (STZ) is widely used to induce
Prasenjit Manna et al.
Archives of toxicology, 82(3), 137-149 (2008-01-17)
Arsenic, one of the most harmful metalloids, is ubiquitous in the environment. The present study has been carried out to investigate the protective role of a triterpenoid saponin, arjunolic acid (AA) against arsenic-induced cardiac oxidative damage. In the study, NaAsO2
Jyotirmoy Ghosh et al.
Biochimie, 95(6), 1098-1109 (2013-02-14)
In recent years, a number of studies describing the effective therapeutic strategies of medicinal plants and their active constituents in traditional medicine have been reported. Indeed, tremendous demand for the development and implementation of these plant derived biomolecules in complementary
Rudiyansyah et al.
Journal of natural products, 69(8), 1218-1221 (2006-08-29)
Phytochemical exploration of a wood bark extract from Durio zibethinus afforded two new triterpenoids, namely, methyl 27-O-trans-caffeoylcylicodiscate (1) and methyl 27-O-cis-caffeoylcylicodiscate (2), a new phenolic, 1,2-diarylpropane-3-ol (3), and seven known compounds, fraxidin, eucryphin, boehmenan, threo-carolignan E, (-)-(3R,4S)-4-hydroxymellein, methyl protocatechuate, and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service