Skip to Content
Merck
All Photos(1)

Key Documents

SMB00083

Sigma-Aldrich

Sweroside

≥95% (LC/MS-ELSD)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H22O9
CAS Number:
Molecular Weight:
358.34
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.25

Assay

≥95% (LC/MS-ELSD)

form

solid

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@@H](O[C@@H]2OC=C3[C@@H](CCOC3=O)[C@H]2C=C)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C16H22O9/c1-2-7-8-3-4-22-14(21)9(8)6-23-15(7)25-16-13(20)12(19)11(18)10(5-17)24-16/h2,6-8,10-13,15-20H,1,3-5H2/t7-,8+,10-,11-,12+,13-,15+,16+/m1/s1

InChI key

VSJGJMKGNMDJCI-ZASXJUAOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Natural product derived from plant source.
Sweroside is an active component of iridoid glycoside, with a molecular weight of 358.344 Da. It is found in variety of plant species.

Application

Sweroside has been used as a standard in MC-high performance liquid chromatography (HPLC)-ultraviolet (UV) experiment.

Biochem/physiol Actions

Sweroside possesses hepatoprotective, antitumor, antibacterial and antifungal properties. It can be used as a potential therapeutic to treat osteoporosis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhong-Duo L Yang et al.
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica, 33(3), 269-272 (2008-06-10)
To study the chemical constituents of the root of Hemerocallis fulva. Compounds were isolated by repeated silica gel, Sephadex LH-20, MCI gel, etc. column chromatography and their structures were determined by spectral analyses and physicochemical properties. From the n-butanol fraction
M M Horn et al.
Phytochemistry, 57(1), 51-56 (2001-05-05)
From the rootstock of Scabiosa columbria L. loganin and sweroside were isolated. Sweroside showed moderate antibacterial activity. Pure sweroside was converted to the sweroside aglycone 1-acetoxy derivative (DABCO/Ac2O) after hydrolysis of the glucose unit. X-ray crystallography of the monoacetate showed
Anne T Stevens et al.
Organic & biomolecular chemistry, 7(17), 3527-3536 (2009-08-14)
A Diels-Alder cycloaddition approach to the sweroside aglycone intermediate of iridoids was explored using silylated butenolides and levoglucosenone as dienophiles under both Lewis acid and thermal conditions. Results of this study reveal no evidence that using less sterically demanding derivatives
Hui Sun et al.
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica, 33(17), 2161-2164 (2008-12-11)
To elucidate the promoting effect of the constituents in plasma after oral administration of Liuwei Dihuangwan on proliferation of rat osteoblast. To add the constituents in plasma after oral administration of Liuwei Dihuangwan with rational concentration into culture solution of
Satyendra Suryawanshi et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 858(1-2), 211-219 (2007-09-18)
Extracts from Swertia chirata (family Gentianaceae) have antidiabetics and antioxidant activity, largely attributed to the flavonoids and secoiridoids, which are a major class of functional components in methanolic extracts from aerial part of plants. In order to facilitate analysis of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service