Skip to Content
Merck
All Photos(1)

Key Documents

SCP0260

Sigma-Aldrich

[Ac-Tyr1,D-Phe2]-VIP Antagonist-GRF 1-29

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C157H252N44O43S1
Molecular Weight:
3476.01
UNSPSC Code:
12352209
NACRES:
NA.32

Assay

≥95% (HPLC)

form

lyophilized

composition

Peptide Content, ≥75%

storage condition

protect from light

storage temp.

−20°C

Amino Acid Sequence

Ac-Tyr-Phe-Asp-Ala-Ile-Phe-Thr-Asn-Ser-Tyr-Arg-Lys-Val-Leu-Gly-Gln-Leu-Ser-Ala-Arg-Lys-Leu-Leu-Gln-Asp-Ile-Met-Ser-Arg-NH2

Application

[Ac-Tyr1,D-Phe2]-GHRF(1-29); [Ac-Tyr1,D-Phe2]-GRF(1-29) is used as an antagonist of vasoactive intestinal peptide (VIP) via its receptor, VPAC.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kai Yang et al.
Hippocampus, 19(9), 779-789 (2009-01-29)
Vasoactive intestinal peptide (VIP) is a 28-amino acid peptide, which belongs to a superfamily of structurally related peptide hormones including pituitary adenylate cyclase-activating polypeptide (PACAP). Although several studies have identified the involvement of PACAP in learning and memory, little work
Jason Itri et al.
Journal of neurophysiology, 90(3), 1589-1597 (2003-09-11)
Circadian rhythmicity in mammals is generated by a pair of nuclei in the anterior hypothalamus known as the suprachiasmatic nuclei (SCN), whose neurons express a variety of neuropeptides that are thought to play an important role in the circadian timing
J Corbitt et al.
Neuropeptides, 36(1), 34-45 (2002-07-31)
VIP and PACAP38 are closely related peptides that are released in the adrenal gland and sympathetic ganglia and regulate catecholamine synthesis and release. We used PC12 cells as a model system to examine receptor and second messenger pathways by which
D M Liu et al.
The European journal of neuroscience, 12(7), 2243-2251 (2000-08-18)
The effects of vasoactive intestinal polypeptide (VIP) and pituitary adenylate cyclase-activating polypeptide (PACAP27 and PACAP38) on isolated parasympathetic neurons of rat intracardiac and submandibular ganglia were examined under voltage clamp using whole-cell patch-clamp recording techniques. VIP and PACAP (</= 10
Tejashree Redij et al.
Chemical biology & drug design, 99(6), 857-867 (2022-03-22)
We report the discovery of two new 2-aminothiophene based small molecule positive allosteric modulators (PAMs) of glucagon-like peptide 1 receptor (GLP-1R) for the treatment of type 2 diabetes. One of the chemotypes, (S-1), has a molecular weight of 239 g/mol

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service