SCP0093
Caspase Inhibitor
≥95% (HPLC)
Synonym(s):
Ac-LETD-CHO, caspase 8 inhibitor, caspase 9 inhibitor
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C21H34N4O10
Molecular Weight:
502.52
UNSPSC Code:
12352200
NACRES:
NA.32
Recommended Products
Assay
≥95% (HPLC)
form
lyophilized
composition
Peptide Content, ≥80%
storage condition
protect from light
storage temp.
−20°C
Gene Information
human ... CASP8(841) , CASP9(842)
Amino Acid Sequence
Ac-Leu-Glu-Thr-Asp-al
Application
Caspase inhibitor Ac-LETD-CHO has been used to inhibit caspase-8.
Biochem/physiol Actions
The peptide Ac-Leu-Glu-Thr-Asp-al, or Ac-LETD-CHO, is a known caspase inhibitor. Ac-LETD-CHO has been reported to exert inhibitory effects on caspase 8 and on caspase 9.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Preventive Effects of Food Components on Caspase-8-Mediated Apoptosis Induced by Dietary Carcinogen, Trp-P-1, in Rat Mononuclear Cells
Hashimoto, T., et al.
ACS Symp. Ser., 851, 128-140 (2003)
Jie Shi et al.
Biochemical pharmacology, 75(9), 1729-1742 (2008-04-02)
Indirubin-3'-monoxime (I3M) is a derivative of indirubin, an active component from a Chinese medicinal recipe with known anti-cancer function. I3M has been well established as a cyclin-dependent kinase (CDK) inhibitor, while the molecular mechanism underlying I3M-induced apoptosis has not been
Apoptotic Regulation of Caspase Activity
Martinova, E.A., et al.
Russian J. Bioorg. Chem., 29, 471-495 (2003)
Enteric glial cells counteract Clostridium difficile Toxin B through a NADPH oxidase/ROS/JNK/caspase-3 axis, without involving mitochondrial pathways
Lara M
Scientific Reports (2017)
Takashi Araki et al.
Nephron. Experimental nephrology, 93(3), e117-e124 (2003-03-28)
Programmed cell death is a mechanism by which organisms dispose of unwanted cells, and it is thought to be an important process in organogenesis. We have already reported the role of caspase-3 in the developing metanephros. While caspase-3 is thought
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service