SCP0091
Caspase 9 Substrate, chromogenic
≥95% (HPLC), lyophilized
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C29H38N8O11
Molecular Weight:
674.66
UNSPSC Code:
12352202
NACRES:
NA.32
Recommended Products
Product Name
Caspase 9 Substrate, chromogenic, ≥95% (HPLC)
Assay
≥95% (HPLC)
form
lyophilized
composition
Peptide Content, ≥75%
storage condition
protect from light
storage temp.
−20°C
Related Categories
Amino Acid Sequence
Ac-Leu-Glu-His-Asp-pNA
Application
Caspase 9 is an initiator caspase that cleaves procaspase 3 and procaspase 7 to propagate the apoptotic cascade. Caspase 9 may be detected using a variety of chromogenic and fluorogenic peptide substrates built around the LEHD peptide core such as the chromogenic substrate Ac-LEHD-pNa (Ac-Leu-Glu-His-Asp-p-nitroanalide).
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Yinyuan Wu et al.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology, 27(5), 539-546 (2011-06-22)
Oxidative stress is known to be involved in a variety of pathological processes including atherosclerosis, diabetes, and neurodegenerative diseases. Understanding how intracellular signaling pathways respond to oxidative stress will have a significant implication in the therapy of these diseases. In
Lin-Li Zhou et al.
European journal of pharmacology, 670(2-3), 399-408 (2011-09-29)
Psoriasis is a chronic inflammatory skin disease affecting 1-3% of the world's population. Traditional Chinese medicines have been extensively used for treating psoriasis with promising clinical results. Celastrol, a triterpenoid isolated from a Chinese herb Celastrus orbiculatus caulis, has been
Juan Liu et al.
Molecular cancer, 10, 95-95 (2011-08-02)
Caspase-8 is a key upstream mediator in death receptor-mediated apoptosis and also participates in mitochondria-mediated apoptosis via cleavage of proapoptotic Bid. However, the role of caspase-8 in p53- and p73-dependent apoptosis induced by genotoxic drugs remains unclear. We recently reported
Aza-derivatives of resveratrol are potent macrophage migration inhibitory factor inhibitors.
Fujita, Y., et al.
Investigational New Drugs (2011)
Sriram Ravindran et al.
Journal of biochemical and molecular toxicology, 34(12), e22606-e22606 (2020-09-01)
One of the common negative impacts in the management of acute myocardial infarction is cognitive decline. Using the rat model of isoproterenol (ISO)-induced myocardial infarction, we assessed the cardioprotective effect of sodium thiosulfate (STS) and its influence on cognition. STS
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service