SBR00073
Non Polar Metabolites QC Mix
suitable for LC/MS
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41116107
NACRES:
NA.84
Recommended Products
form
liquid
Quality Level
technique(s)
LC/MS: suitable
application(s)
lipidomics
metabolomics
storage temp.
−20°C
Related Categories
General description
In order to assess any variations in the LC-MS-based metabolomics analysis data, it is recommended to add an additional QC (quality control) sample at the beginning of every analytical experiment batch and should be injected every 4-10 injections in to the workflow steps. The QC sample evaluates any drifting phenomenons like intensity values, ion suppression or any changes in the retention time of the materials peaks in the LC-MS data.
The Non Polar Metabolites QC Mix is a ready to use solution (~2:1:1 IPA:ACN:H20) for mass spectroscopy workflow. By using the QC mix, the user will be able to assess drifting and ion suppresion phenomenons. The Non Polar Metabolites QC Mix comprised from nine components solution mix which contains a hydrophobic type of metabolites such as, cholesterol derivatives, unsaturated fatty acids, phospholipids and ceramides.
The Non Polar Metabolites QC Mix is a ready to use solution (~2:1:1 IPA:ACN:H20) for mass spectroscopy workflow. By using the QC mix, the user will be able to assess drifting and ion suppresion phenomenons. The Non Polar Metabolites QC Mix comprised from nine components solution mix which contains a hydrophobic type of metabolites such as, cholesterol derivatives, unsaturated fatty acids, phospholipids and ceramides.
Features and Benefits
- The non polar metabolites QC mix covers ~ 100-800 Da mass range
- The mix was designed for ESI (+/-) detection
- The mixture comes as a ready to use solution (~2:1:1 IPA:ACN:H20) format in crimp top (Silicone/PTFE liner) amber vial which suitable for use with most of the LC-MS autosamplers
- The recommended storage temperature of the mixture is -20°C, thawing of the sample is not required. Warm the sample to room temparature and mix briefly
Components
Progesterone
D-Sphingosine
1-Oleoyl-sn-glycero-3-phosphocholine (LysoPC(18:1(9Z)/0:0))
Linolenic acid
Arachidonic acid sodium salt
cis-4,7,10,13,16,19-Docosahexaenoic acid
Cholesterol sulfate
N-palmitoyl-D-sphingosine (C16 Ceramide (d18:1/16:0))
1,2-Dioleoyl-sn-glycero-3-phosphocholine (18:1 (Δ9-Cis) PC (DOPC))
D-Sphingosine
1-Oleoyl-sn-glycero-3-phosphocholine (LysoPC(18:1(9Z)/0:0))
Linolenic acid
Arachidonic acid sodium salt
cis-4,7,10,13,16,19-Docosahexaenoic acid
Cholesterol sulfate
N-palmitoyl-D-sphingosine (C16 Ceramide (d18:1/16:0))
1,2-Dioleoyl-sn-glycero-3-phosphocholine (18:1 (Δ9-Cis) PC (DOPC))
related product
Product No.
Description
Pricing
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 3 - STOT SE 3
Target Organs
Central nervous system
Storage Class Code
3 - Flammable liquids
WGK
WGK 2
Flash Point(F)
89.6 °F
Flash Point(C)
32 °C
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service