Skip to Content
Merck
All Photos(3)

Key Documents

SAB5200022

Sigma-Aldrich

Monoclonal Anti-CACNA1D antibody produced in mouse

clone S48A-9, 1 mg/mL, purified immunoglobulin

Synonym(s):

Anti-CACH3, Anti-CACN4, Anti-CACNA 1D, Anti-CACNL1A2, Anti-Cav1.3, S48A-9, Anti-oltage dependent L type calcium channel subunit alpha 1D

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
Pricing and availability is not currently available.

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

S48A-9, monoclonal

form

buffered aqueous glycerol solution

mol wt

antigen predicted mol wt 250 kDa

species reactivity

human, rat, mouse

concentration

1 mg/mL

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Calcium channel, L type α-1 polypeptide isoform 2 (CACNA1D) is expressed in many tissues and possesses six transmembrane segments, along with a loop lining the pore of the channel. The gene encoding CACNA1D is localized on human chromosome 3p14.3.

Specificity

Detects ~250 kDa. No cross-reactivity against Cav1.2

Immunogen

Fusion protein amino acids 859-875 of rat Cav1.3, Accession Number P27732

Application

Monoclonal Anti-CACNA1D antibody produced in mouse has been used in immunocytochemistry.[1]

Biochem/physiol Actions

Calcium channel, L type α-1 polypeptide isoform 2 (CACNA1D) is crucial for the functioning of the brain and in hearing. It gets activated at a fast rate and withstands high inward Ca2+ currents even at threshold voltages. CACNA1D aids in dendritic signals linked to mitochondrial stress in the brain and is important for estrogen-stimulated Ca2+ influx. Studies have shown that in atrial myocytes, the C terminus of the channel translocates to the nucleus and regulates Ca2+-activated K+ channels. CACNA1D was shown to be upregulated in endometrial cancer and its knockdown showed a reduction in cell proliferation in vitro. Mutations in the gene encoding it has been associated with aldosterone-producing adenomas (APAs), where it leads to the formation of small adenomas.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in PBS, pH 7.4, 50% glycerol, and 0.09% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juan Hao et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 29(7), 2883-2893 (2015-03-26)
Calcium and calcium channels are closely related to the estrogen-induced nongenomic effect of endometrial carcinoma, but the specific role of calcium channels is unknown. This study aimed to explore the expression and the biologic effect of the L-type calcium channel
Andreas Lieb et al.
Biophysical journal, 106(7), 1467-1475 (2014-04-08)
Activity of voltage-gated Cav1.3 L-type Ca(2+) channels is required for proper hearing as well as sinoatrial node and brain function. This critically depends on their negative activation voltage range, which is further fine-tuned by alternative splicing. Shorter variants miss a
Fabio Luiz Fernandes-Rosa et al.
Hypertension (Dallas, Tex. : 1979), 64(2), 354-361 (2014-05-29)
Primary aldosteronism is the most common form of secondary hypertension. Somatic mutations in KCNJ5, ATP1A1, ATP2B3, and CACNA1D have been described in aldosterone-producing adenomas (APAs). Our aim was to investigate the prevalence of somatic mutations in these genes in unselected
Assignment of the gene encoding the alpha 1 subunit of the neuroendocrine/brain-type calcium channel (CACNL1A2) to human chromosome 3, band p14.3.
S Seino et al.
Genomics, 13(4), 1375-1377 (1992-08-01)
Ling Lu et al.
The Journal of biological chemistry, 290(8), 4663-4676 (2014-12-30)
Cav1.3 L-type Ca(2+) channel is known to be highly expressed in neurons and neuroendocrine cells. However, we have previously demonstrated that the Cav1.3 channel is also expressed in atria and pacemaking cells in the heart. The significance of the tissue-specific

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service