Skip to Content
Merck
All Photos(1)

Key Documents

SAB4700731

Sigma-Aldrich

Monoclonal Anti-HLA-DR antibody produced in mouse

clone L243, purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

L243, monoclonal

form

buffered aqueous solution

species reactivity

nonhuman primates, human, dog

concentration

1 mg/mL

technique(s)

FUNC: suitable
flow cytometry: suitable
immunoblotting: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable

isotype

IgG2a

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... HLA-DRA(3122)

General description

Human leukocyte antigen (HLA)-DR is a member of HLA class II molecules. It is encoded by the gene mapped to human chromosome 6.(21} HLA class II molecules are highly expressed on B-Iymphocytes, but is also found at minimal levels on T-Iymphocytes, macrophages, thymus epithelial cells and selected cells of various body tissues. HLA-DR consists of a non-polymorphic α chain gene and up to three distinct highly polymorphic β chain genes.
The mouse monoclonal antibody L243 recognizes specifically HLA-DR molecules, both peptide-loaded and empty.

Immunogen

Human B lymphocytes

Application

Monoclonal Anti-HLA-DR antibody produced in mouse has been used in double immunofluorescent staining.

Biochem/physiol Actions

Human leukocyte antigen (HLA)-DR activates a T-cell receptor by presenting antigenic peptides derived from extracellular proteins thus, stimulating an immune response against the antigen.{22] Mutation in the gene increases the risk of susceptibility to inflammatory bowel disease (IBD), Crohn′s disease and ulcerative colitis. Overexpression of monocytic (HLA)-DR has been observed in schizophrenic patients.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4, with 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reactive microglia in patients with senile dementia of the Alzheimer type are positive for the histocompatibility glycoprotein HLA-DR.
McGeer PL, et al.
Neuroscience Letters, 79(1-2), 195-200 (1987)
HLA-DR and -DQ phenotypes in inflammatory bowel disease: a meta-analysis
Stokkers PC, et al.
Gut, 45(3), 395-401 (1999)
Monocytic HLA DR antigens in schizophrenic patients.
Krause D, et al.
Neuroscience Research, 72(1), 87-93 (2012)
Expression of pannexin 1 and 2 in cortical lesions from intractable epilepsy patients with focal cortical dysplasia
Li S, et al.
Oncotarget, 8(4), 6883-6883 (2017)
Eric Alix et al.
Cell host & microbe, 28(1), 54-68 (2020-06-12)
The Salmonella enterica effector SteD depletes mature MHC class II (mMHCII) molecules from the surface of infected antigen-presenting cells through ubiquitination of the cytoplasmic tail of the mMHCII β chain. Here, through a genome-wide mutant screen of human antigen-presenting cells

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service