Skip to Content
Merck
All Photos(1)

Key Documents

SAB4700563

Sigma-Aldrich

Anti-Cd8a low endotoxin antibody, Rat monoclonal

clone 53-6.7, purified immunoglobulin, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rat

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

53-6.7, monoclonal

form

buffered aqueous solution

species reactivity

mouse

concentration

1 mg/mL

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunohistochemistry: suitable
immunoprecipitation (IP): suitable

impurities

≤1 EU/μg endotoxin (LAL)

isotype

IgG2a

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

mouse ... Cd86(12524)

General description

The rat monoclonal antibody 53-6.7 recognizes mouse CD8a (32-34 kDa; alpha chain of the CD8 antigen).

Immunogen

Mouse spleen cells

Application

The reagent is designed for Flow Cytometry analysis. Suggested working dilution is 1.5 μg/mL of sample. Indicated dilution is recommended starting point for use of this product. Working concentrations should be determined by the investigator.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in azide free phosphate buffered saline, pH 7.4; 0.2 um filter sterilized. Endotoxin level is less than 0.01 EU/μg of the protein, as determined by the LAL
test.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Scott A Gerber et al.
International journal of cancer, 134(10), 2383-2392 (2013-10-25)
Radiation therapy (RT) continues to be a cornerstone in the treatment for many cancers. Unfortunately, not all individuals respond effectively to RT resulting clinically in two groups consisting of nonresponders (progressive disease) and responders (tumor control/cure). The mechanisms that govern
P McGuirk et al.
European journal of immunology, 28(1), 153-163 (1998-03-04)
We have used a murine respiratory challenge model to examine the local T cell responses in the lung during infection with Bordetella pertussis. T cells from lung parenchyma and airways of naive and infected mice were refractory to both antigen
M J Smyth et al.
Journal of virology, 72(7), 5948-5954 (1998-06-17)
Mouse cytotoxic T lymphocytes (CTL) reactive with a H-2Db-presented 9-mer peptide of the human papillomavirus type 16 protein E7(49-57) (RAHYNIVTF) were generated from the spleen cells of wild-type C57BL/6 (B6) or B6 perforin-deficient (B6.P0) mice. CD8(+) B6 CTL displayed peptide-specific
G Das et al.
The Journal of experimental medicine, 190(6), 881-884 (1999-09-28)
Peripheral CD8(+) T cells mainly use CD8alpha/beta, and their development is mainly dependent on the major histocompatibility complex (MHC) class I proteins K(b) and D(b) in H-2(b) mice. In this report, we have shown that the development of CD8alpha/beta TCR-alpha/beta
S Stäger et al.
Journal of immunology (Baltimore, Md. : 1950), 165(12), 7064-7071 (2000-12-20)
Vaccination against visceral leishmaniasis has received limited attention compared with cutaneous leishmaniasis, although the need for an effective vaccine against visceral leishmaniasis is pressing. In this study, we demonstrate for the first time that a recombinant stage-specific hydrophilic surface protein

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service