Skip to Content
Merck
All Photos(10)

Documents

SAB4504331

Sigma-Aldrich

Anti-phospho-Akt (pSer473) antibody produced in rabbit

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 55 kDa

species reactivity

human, mouse, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:5000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer473)

Gene Information

human ... AKT1(207)
rat ... Akt1(24185)

General description

AKT1 (AKT serine/threonine kinase 1) is also called as v-akt murine thymoma viral oncogene homolog 1 and PKBα (protein kinase B). It is expressed in liver, muscle and adipocytes. AKT1 gene is mapped to human chromosome 14q32. It is a member of AKT family that has an amino-terminal pleckstrin homology (PH) domain, a short α helical linker and a carboxyl-terminal kinase domain.

Immunogen

The antiserum was produced against synthesized peptide derived from human Akt around the phosphorylation site of Ser473.

Immunogen Range: 431-480

Application

Anti-phospho-Akt (pSer473) antibody has been used in western blotting.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

AKT1 (AKT serine/threonine kinase 1) controls the survival of cells and anti-apoptotic actions that attack the pathogenesis of several cancers , hence it plays a crucial role in tumorigenesis. In mice, AKT1 is essential for normal development.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Association between single nucleotide polymorphisms in AKT1 and the risk of prostate cancer in the Chinese Han population
Liu JM, et al.
Genetics and molecular research : GMR, 16(1) (2017)
Shih-Ron Hsieh et al.
Journal of biomedical science, 20, 86-86 (2013-11-21)
Epigallocatechin-3-gallate (EGCg) with its potent anti-oxidative capabilities is known for its beneficial effects ameliorating oxidative injury to cardiac cells. Although studies have provided convincing evidence to support the cardioprotective effects of EGCg, it remains unclear whether EGCg affect trans-membrane signalling
J Matthew Kuczmarski et al.
Experimental physiology, 103(4), 545-558 (2018-01-10)
What is the central question of this study? Translocation of nNOSμ initiates catabolic signalling via FoxO3a and skeletal muscle atrophy during mechanical unloading. Recent evidence suggests that unloading-induced muscle atrophy and FoxO3a activation are redox sensitive. Will a mimetic of
Epigallocatechin-3-gallate-mediated cardioprotection by Akt/GSK-3?/caveolin signalling in H9c2 rat cardiomyoblasts
Hsieh SR, et al.
Journal of Biomedical Science (2013)
Amit Joharapurkar et al.
Basic & clinical pharmacology & toxicology, 130(1), 35-43 (2021-10-12)
Inhibiting the intestinal and renal neutral amino acid transporter B0AT1 by genetic means has improved insulin sensitivity in mice, but there are no antagonists available for preclinical or clinical use. Since the anti-inflammatory agent nimesulide selectively inhibited B0AT1 in vitro

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service