Skip to Content
Merck
All Photos(2)

Key Documents

SAB4503853

Sigma-Aldrich

Anti-phospho-Akt (pSer124) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-AKT, Anti-PKB, Anti-PKB-ALPHA, Anti-PRKBA, Anti-RAC, Anti-RAC-ALPHA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 55 kDa

species reactivity

human, mouse, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:5000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer124)

Gene Information

human ... AKT1(207)

Immunogen

The antiserum was produced against synthesized peptide derived from human Akt around the phosphorylation site of Ser124.

Immunogen Range: 90-139

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guanxue Xu et al.
Bioscience reports, 40(5) (2020-05-13)
Matrine is a main active constituent of Chinese herb Sophora flavescens Ait (Kushen), which has shown various pharmacological effects, and has been reported to exhibit protective effects in heart failure. In the present study, the underlying mechanism of matrine was
L Zhang et al.
Neoplasma, 67(4), 762-772 (2020-07-30)
Malignant glioma is the most frequent primary brain tumor in adults. Accumulated evidence showed that long non-coding RNA (lncRNA) long intergenic noncoding RNA 152 (LINC00152) participated in the progression of glioma, while the regulatory mechanisms remain elusive. Here, the study
Xiuli Zhang et al.
International journal of molecular medicine, 35(6), 1747-1754 (2015-04-16)
Zinc (Zn) as an essential dietary element has been indicated in a number of protein functions in the prevention of numerous types of epithelial-to-mesenchymal transition (EMT)-driven fibrosis in vivo. However, relatively little is known regarding its effect in the EMT
Lili Liu et al.
Bioscience reports, 40(5) (2020-05-19)
The present study aims to reveal the molecular mechanism of peroxisome proliferator-activated receptor γ (PPARγ) on sepsis-induced acute lung injury (ALI). To do that, the rat injury model was established using cecal ligation and perforation (CLP) method, followed by different
Ming Tu et al.
Molecular medicine reports, 15(4), 1713-1721 (2017-03-06)
Cluster of differentiation 164 (CD164), a sialomucin, has been demonstrated to be involved in the regulation of proliferation, apoptosis, adhesion and differentiation in multiple cancers. CD164 is regarded to be a potential promotor of tumor growth. However, the involvement of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service