Skip to Content
Merck
All Photos(1)

Key Documents

SAB4502829

Sigma-Aldrich

Anti-Synuclein-α antibody produced in rabbit

affinity isolated antibody

Synonym(s):

α-synuclein, NACP, Non-A β component of AD amyloid, Non-A4 component of amyloid precursor, SNCA

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 14 kDa

species reactivity

mouse, rat, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:40000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SNCA(6622)

General description

Anti-Synuclein-α antibody detects endogenous levels of total Synuclein-α protein.
The SNCA (synuclein α) gene is mapped to human chromosome 4q22.1.

Immunogen

The antiserum was produced against synthesized peptide derived from human Synuclein-alpha.

Immunogen Range: 86-135

Biochem/physiol Actions

Accumulation of α-synuclein in oligodendrocytes results in the loss of myelin and causes neurodegeneration, leading to multiple system atrophy. Synuclein-α is known to induce oligodendrocyte maturation. The encoded protein is considered to be an important aggregate of Lewy bodies, contributing to the pathogenesis of Parkinson disease.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jian-Feng Li et al.
Foods (Basel, Switzerland), 13(9) (2024-05-11)
Parkinson's disease (PD), the second most common neurodegenerative disorder, is linked to α-synuclein (α-Syn) aggregation. Despite no specific drug being available for its treatment, curcumin, from the spice turmeric, shows promise. However, its application in PD is limited by a
SNCA variants are associated with increased risk for multiple system atrophy.
Scholz SW
Annals of Neurology, 65(5), 610-614 (2009)
?-Synuclein-induced myelination deficit defines a novel interventional target for multiple system atrophy.
Ettle B
Acta Neuropathologica, 132(1), 59-75 (2016)
Synucleins Have Multiple Effects on Presynaptic Architecture.
Vargas KJ
Cell Reports, 18(1), 161-173 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service