Skip to Content
Merck
All Photos(1)

Key Documents

SAB4502101

Sigma-Aldrich

Anti-TNNI3K antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Cardiac ankyrin repeat kinase TNNI3K, Serine/threonine-protein kinase TNNI3K TNNI3-interacting kinase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 104 kDa

species reactivity

rat, mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
immunohistochemistry: 1:50-1:100

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TNNI3K(51086)

General description

Anti-TNNI3K antibody detects endogenous levels of total TNNI3K protein.
The TNNI3K (TNNI3 interacting kinase) gene is a novel cardiac troponin I-interacting kinase gene and is mapped to human chromosome 1p31.1. The gene is expressed in cardiomyocytes. The encoded protein belongs to mitogen-activated protein kinase kinase kinase (MAPKKK) family. Tnni3k consists of seven to ten ankyrin repeats at the N terminal, a Ser/Thr and Tyr kinase domain, and a Ser-rich domain at the C-terminal.

Immunogen

The antiserum was produced against synthesized peptide derived from human TNNI3K.

Immunogen Range: 301-350

Biochem/physiol Actions

Overexpression of TNNI3K (TNNI3 interacting kinase) may induce and increase cardiac myogenesis and performance respectively. It also might inhibit ventricular remodeling, caused by ischemia. TNNI3K might be associated with the development of cardiac hypertrophy. TNNI3K is known to be associated with atrioventricular conduction disease, acute coronary syndrome and obesity.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Overexpression of Cardiac-Specific Kinase TNNI3K Promotes Mouse Embryonic Stem Cells Differentiation into Cardiomyocytes.
Wang Y
Cellular Physiology and Biochemistry, 41(1), 381-398 (2017)
TNNI3K could be a novel molecular target for the treatment of cardiac diseases.
Lai ZF
Recent Patents on Cardiovascular Drug Discovery, 4(3), 203-210 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service