Skip to Content
Merck
All Photos(3)

Key Documents

SAB4501954

Sigma-Aldrich

Anti-HNRNP M antibody produced in rabbit

affinity isolated antibody

Synonym(s):

HNRPM, heterogeneous nuclear ribonucleoprotein M

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 77 kDa

species reactivity

human, rat, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:40000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... HNRNPM(4670)

General description

Anti-HNRNP M Antibody detects endogenous levels of total HNRNP M protein.
HNRNP M (heterogeneous nuclear ribonucleoprotein M) gene is mapped to human chromosome 19p13.2. It codes for a nucleocytoplasmic shuttling RNA binding protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human hnRNP M.

Immunogen Range: 11-60

Biochem/physiol Actions

HnRNP M (heterogeneous nuclear ribonucleoprotein M) is an RNA binding protein and participates in mRNA splicing mechanism. It is significantly involved in the formation of mature mRNAs from heterogeneous nuclear RNAs. The encoded protein acts as an important gene expression regulating factor. HnRNPM is involved in the activation of IRES (internal ribosomal entry site)-dependent translation of FGF1 (fibroblast growth factor 1). Thus, this protein is associated with myoblast differentiation. HNRNP M expression is associated with the invasion and metastasis events of tumor cells. Upregulation of HNRNP M is observed in breast cancer and human colorectal epithelial carcinogenesis. Therefore, HnRNP M may serve as an important biomarker for cancer.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shuijiao Chen et al.
American journal of physiology. Gastrointestinal and liver physiology, 306(5), G394-G403 (2014-01-02)
Colorectal carcinoma (CRC) is one of the most common cancers in the world, and identification of new CRC biomarkers will be helpful for the diagnosis and treatment of CRC. For isobaric tags for relative and absolute quantitation (iTRAQ) analysis, fresh
Huizhi Sun et al.
Genes, chromosomes & cancer, 56(8), 598-607 (2017-04-11)
HnRNPM is an essential splicing factor and its expression is closely correlated with invasion and metastasis of tumor cells. The CD44 cell adhesion molecule is aberrantly expressed in many breast tumors and CD44 splice variants have been implicated in specific
Jacqueline Smith et al.
Mammalian genome : official journal of the International Mammalian Genome Society, 13(6), 310-315 (2002-07-13)
Human Chromosome 19 (HSA19) is virtually completely sequenced. A complete physical contig map made up of BACs and cosmids is also available for this chromosome. It is, therefore, a rich source of information that we have used as the basis
Julienne M Jagdeo et al.
Journal of virology, 89(14), 7064-7078 (2015-05-01)
Picornavirus infection involves a dynamic interplay of host and viral protein interactions that modulates cellular processes to facilitate virus infection and evade host antiviral defenses. Here, using a proteomics-based approach known as TAILS to identify protease-generated neo-N-terminal peptides, we identify
Jun-ichi Takino et al.
World journal of gastroenterology, 21(6), 1784-1793 (2015-02-17)
To study the formation of intracellular glyceraldehyde-derived advanced glycation end products (Glycer-AGEs) in the presence of high concentrations of fructose. Cells of the human hepatocyte cell line Hep3B were incubated with or without fructose for five days, and the corresponding

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service