Skip to Content
Merck
All Photos(2)

Documents

SAB4501315

Sigma-Aldrich

Anti-ASC antibody produced in rabbit

affinity isolated antibody

Synonym(s):

ASC, CARD5, apoptosis-associated speck-like protein containing a CARD, caspase recruitment domain protein 5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 21 kDa

species reactivity

rat, human, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:40000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PYCARD(29108)

General description

Anti-ASC Antibody detects endogenous levels of total ASC protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human ASC.

Immunogen Range: 10-59

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jin-Ling Ren et al.
Cell death & disease, 12(5), 436-436 (2021-05-03)
Atherosclerotic plaque vulnerability and rupture increase the risk of acute coronary syndromes. Advanced lesion macrophage apoptosis plays important role in the rupture of atherosclerotic plaque, and endoplasmic reticulum stress (ERS) has been proved to be a key mechanism of macrophage
Jin Ye et al.
Stem cell research & therapy, 15(1), 236-236 (2024-07-30)
Abnormalities in T cell activation play an important role in the pathogenesis of myocarditis, and persistent T cell responses can lead to autoimmunity and chronic cardiac inflammation, as well as even dilated cardiomyopathy. Although previous work has examined the role
Long-Yun Zhou et al.
Journal of neurochemistry, 155(2), 154-176 (2020-03-28)
Cervical spondylotic myelopathy (CSM) is a common cause of disability with few treatments. Aberrant mitochondrial dynamics play a crucial role in the pathogenesis of various neurodegenerative diseases. Thus, regulation of mitochondrial dynamics may offer therapeutic benefit for the treatment of
Hui Liu et al.
The Journal of pathology, 242(3), 284-296 (2017-04-06)
Acute-phase inflammation plays a critical role in liver graft injury. Inflammasomes, multi-molecular complexes in the cytoplasm, are responsible for initiating inflammation. Here, we aimed to explore the role of inflammasomes in liver graft injury and further to investigate the regulatory
Chao Ma et al.
Human & experimental toxicology, 41, 9603271211061497-9603271211061497 (2022-02-22)
As a common postoperative complication to elderly patients, postoperative cognitive dysfunction (POCD) is a central nervous system complication, often taking place after anesthesia and surgery. (Su(var)3-9, enhancer-of-zeste, and trithorax) domain-containing protein 7 (SETD7) plays important roles in metabolic-related diseases, viral

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service