Skip to Content
Merck
All Photos(3)

Key Documents

SAB4501162

Sigma-Aldrich

Anti-Glial Fibrillary Acidic Protein Antibody

rabbit polyclonal

Synonym(s):

Glial fibrillary acidic protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

product name

Anti-GFAP antibody produced in rabbit, affinity isolated antibody

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 49 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GFAP(2670)

General description

Anti-GFAP Antibody detects endogenous levels of total GFAP protein.
Glial fibrillary acidic protein (GFAP) is an intermediate-filament protein found in the astrocytes of CNS (central nervous system). It is located on human chromosome 17q21.

Immunogen

The antiserum was produced against synthesized peptide derived from human GFAP.

Immunogen Range: 11-60

Application

Anti-GFAP, N-Terminal antibody has been used in immunocytochemical analysis , confocal and fluorescent microscopic imaging , immunostaining and western blotting.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

Glial fibrillary acidic protein (GFAP) regulates intermediate filament network dynamics, cell morphology and focal adhesions.It helps in maintaining myelination and stability of CNS (central nervous system) white matter architecture.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Childhood constipation; an overview of genetic studies and associated syndromes
Peeters B, et al.
Best Practice & Research. Clinical Gastroenterology, 25 (2011)
MANF Promotes Differentiation and Migration of Neural Progenitor Cells with Potential Neural Regenerative Effects in Stroke
Tseng KY, et al.
Molecular Therapy (2017)
GFAP promoter directs astrocyte-specific expression in transgenic mice
Brenner M, et al.
The Journal of Neuroscience (1994)
Suppression of agrin-22 production and synaptic dysfunction in Cln1 (-/-) mice
Peng S, et al.
Annals of Clinical and Translational Neurology, 2, 1085-1104 (2015)
Ayat I Samra et al.
Biomedicines, 11(12) (2023-12-23)
Autism spectrum disorder (ASD) prevalence is emerging with an unclear etiology, hindering effective therapeutic interventions. Recent studies suggest potential renin-angiotensin system (RAS) alterations in different neurological pathologies. However, its implications in ASD are unexplored. This research fulfills the critical gap

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service