Skip to Content
Merck
All Photos(1)

Documents

SAB4301270

Sigma-Aldrich

Anti-phospho-Nuclear Receptor NR4A1 (pSer351) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

GFRP1, HMR, NAK1, NGFIB, NR4A1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

64 kDa

species reactivity

rat, human, mouse

concentration

1.0 mg/mL

technique(s)

immunohistochemistry: 1:50-1:100

isotype

IgG

accession no.

NP_002126.2.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pSer351)

Gene Information

human ... NR4A1(3164)

General description

Nuclear receptor subfamily 4 group A member 1 (NR4A1) also known as nuclear hormone receptor 77 (NUR77), belongs to NR4A subfamily nuclear hormone receptors. In human chromosome, the gene NR4A1 is localized on 12q13.13.

Specificity

The antibody detects endogenous levels of Nuclear Receptor NR4A1 only when phosphorylated at serine 351

Immunogen

Peptide sequence around phosphorylation site of Serine351(L-P-S(p)-K-P) derived from Human Nuclear Receptor NR4A1.

Biochem/physiol Actions

Nuclear receptor subfamily 4 group A member 1 (NR4A1) is essential for regulation of cell proliferation, apoptosis and energy metabolism. Nur77 is an acetylated protein, modulated by histone acetyltransferase p300 (p300) and histone deacetylase 1 (HDAC1) for its rapid turnover. Nur77 regulates liver kinase B1 (LKB1)- AMP (adenosine monophosphate)-activated protein kinase (AMPK) and plays a major role in glucose metabolism. Expression of Nur77 in skeletal muscle promotes glucose uptake by enhancing insulin pathway and upregulates glycolysis pathway. Expression of Nur77 in hepatocytes promotes gluconeogenesis. Mutation in Nur77 may result in diet-induced obesity and insulin resistance in tissues. Nur77 is a potential drug target for metabolic disorders. NR4A1 is an anti-oncogenic protein that reduces the tumour cell proliferation and metastasis. NR4A1 increases the gastric cancer cells sensitivity towards tumour necrosis factor α (TNF-α) mediated apoptotic pathway. NR4A1 is downregulated in breast cancer, leukemia. NR4A1 plays a crucial role and a potent therapeutic target for fibrosis of endometriosis.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

NR4A1-induced increase in the sensitivity of a human gastric cancer line to TNF alpha-mediated apoptosis is associated with the inhibition of JNK/Parkin-dependent mitophagy
Yan H, et al.
International Journal of Oncology, 52(2), 367-378 (2018)
The orphan nuclear receptor Nur77 regulates LKB1 localization and activates AMPK
Zhan YY, et al.
Nature Chemical Biology, 8(11), 897-904 (2012)
Regulation of Nur77 protein turnover through acetylation and deacetylation induced by p300 and HDAC1
Kang SA, et al.
Biochemical Pharmacology, 80(6), 867-873 (2010)
NR4A1 is Involved in Fibrogenesis in Ovarian Endometriosis
Zeng X, et al.
Cellular Physiology and Biochemistry, 46(3), 1078-1090 (2018)
Merlin G Butler et al.
International journal of molecular sciences, 16(3), 6464-6495 (2015-03-25)
Recently, autism-related research has focused on the identification of various genes and disturbed pathways causing the genetically heterogeneous group of autism spectrum disorders (ASD). The list of autism-related genes has significantly increased due to better awareness with advances in genetic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service