Skip to Content
Merck
All Photos(1)

Key Documents

SAB4300489

Sigma-Aldrich

Anti-KIT (Ab-721) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-C-Kit antibody produced in rabbit, Anti-CD117 antibody produced in rabbit, Anti-PBT antibody produced in rabbit, Anti-SCFR antibody produced in rabbit, Anti-v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~145 kDa

species reactivity

human

concentration

1 mg/mL

technique(s)

western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(N-E-Y-M-D)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... KIT(3815)

Immunogen

Peptide sequence around aa. 719-723 (N-E-Y-M-D), according to the protein KIT.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

This is the receptor for stem cell factor (mast cell growth factor). It has a tyrosine-protein kinase activity. Binding of the ligands leads to the autophosphorylation of KIT and its association with substrates such as phosphatidylinositol 3-kinase (Pi3K).

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marc P Pusztaszeri et al.
Cancer cytopathology, 122(8), 596-603 (2014-06-13)
c-KIT/CD117 down-regulation has been described in papillary thyroid carcinoma (PTC). In this study, the authors investigated CD117 as an ancillary immunocytochemical test for PTC in fine-needle aspiration biopsy (FNAB). The expression of CD117 was assessed in cytologic samples of histologically
Cláudia M Salgado et al.
Pediatric and developmental pathology : the official journal of the Society for Pediatric Pathology and the Paediatric Pathology Society, 17(3), 198-203 (2014-04-01)
Nevocytes (NC) and mastocytes (MC) have different progenitors but share stem cell factor as regulator/activator of NC and for differentiation/proliferation of MC. Both cell types express stem cell factor receptor CD117. We hypothesize that large/giant congenital melanocytic nevi (L/GCMN) may
Francesco Ciccia et al.
Rheumatology (Oxford, England), 53(7), 1313-1320 (2014-03-08)
The aim of this study was to evaluate the role of rituximab (RTX) in modulating the expression of the IL-17/IL-23 pathway in the salivary glands (SGs) of patients with primary SS (pSS). Consecutive SG biopsies were obtained from 15 patients
Sham S Kakar et al.
PloS one, 9(9), e107596-e107596 (2014-09-30)
Currently, the treatment for ovarian cancer entails cytoreductive surgery followed by chemotherapy, mainly, carboplatin combined with paclitaxel. Although this regimen is initially effective in a high percentage of cases, unfortunately within few months of initial treatment, tumor relapse occurs because
Suchandra Chowdhury et al.
Glycoconjugate journal, 31(6-7), 523-535 (2014-10-07)
Childhood acute lymphoblastic leukaemia (ALL) originates from mutations in haematopoietic progenitor cells (HPCs). For high-risk patients, treated with intensified post-remission chemotherapy, haematopoietic stem cell (HSC) transplantation is considered. Autologous HSC transplantation needs improvisation till date. Previous studies established enhanced disease-associated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service