Skip to Content
Merck
All Photos(2)

Key Documents

SAB4300413

Sigma-Aldrich

Anti-HDAC4 (Ab-632) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-HA6116 antibody produced in rabbit, Anti-HD4 antibody produced in rabbit, Anti-HDAC-A antibody produced in rabbit, Anti-HDACA antibody produced in rabbit, Anti-histone deacetylase 4 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~140 kDa

species reactivity

rat, mouse, human

concentration

1 mg/mL

technique(s)

indirect immunofluorescence: 1:100-1:200
western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(A-Q-S-S-P)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... HDAC4(9759)

Immunogen

Peptide sequence around aa. 630-634 (A-Q-S-S-P), according to the protein HDAC4.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Responsible for the deacetylation of lysine residues on the N-terminal part of the core histones (H2A, H2B, H3 and H4). Histone deacetylation gives a tag for epigenetic repression and plays an important role in transcriptional regulation, cell cycle progression and developmental events. Histone deacetylases act via the formation of large multiprotein complexes. Involved in muscle maturation via its interaction with the myocyte enhancer factors such as MEF2A, MEF2C and MEF2D.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Svetlana Demyanenko et al.
Journal of cellular biochemistry, 120(12), 19590-19609 (2019-07-03)
Histone deacetylases (HDAC) inhibitors can protect nerve cells after a stroke, but it is unclear which HDAC isoform is involved in this effect. We studied cellular and intracellular rearrangement of class II HDACs at late periods after photothrombotic infarct (PTI)
Svetlana Demyanenko et al.
International journal of molecular sciences, 20(12) (2019-06-16)
Ischemic penumbra that surrounds a stroke-induced infarction core is potentially salvageable; however, mechanisms of its formation are not well known. Covalent modifications of histones control chromatin conformation, gene expression and protein synthesis. To study epigenetic processes in ischemic penumbra, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service