Skip to Content
Merck
All Photos(2)

Key Documents

SAB4200398

Sigma-Aldrich

Anti-TFRC antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody

Synonym(s):

Anti-CD71, Anti-TFR, Anti-TFR1, Anti-TR, Anti-TRFR, Anti-Transferrin receptor (p90, CD71), Anti-Transferrin receptor protein 1, Anti-p90, AntiT9

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~90 kDa

species reactivity

mouse, rat, human

concentration

~1.0 mg/mL

technique(s)

indirect immunofluorescence: 10-20 μg/mL using rat NRK cells
western blot: 2.5-5.0 μg/mL using whole extracts of human U-2-OS and mouse Neuro-2a cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TFRC(7037)
mouse ... Tfrc(22042)
rat ... Tfrc(64678)

General description

Transferrin receptor (TFRC), also known as p90, CD71, TFR and TFR1, is a transmembrane glycoprotein. It is composed of two disulphide-bonded sub-units, contains three N-linked glycan units and is post-translationally modified with phosphate and fatty acyl groups.

Immunogen

synthetic peptide corresponding to an internal sequence of human TFRC, conjugated to KLH

Application

Anti-TFRC antibody produced in rabbit has been used in immunoblotting and immunofluorescence.

Biochem/physiol Actions

Transferrin receptor (TFRC) mediates cellular iron uptake. It binds iron bound transferrin on the cell surface, and the receptor ligand complex undergoes endocytosis via clathrin coated pits. Endosomal acidification leads to iron release, which is transported to the cytosol or to the mitochondria in erythroid cells. The apotransferrin receptor complex is then recycled to the cell surface, where the affinity of apotransferrin for TFRC drops, resulting in its dissociation. TFRC is necessary for development of erythrocytes and the nervous system.

Physical form

solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

1.6 Mb deletion in chromosome band 3q29 associated with eye abnormalities
Tyshchenko N, et al.
European Journal of Medical Genetics, 52(2-3), 128-130 (2009)
Transferrin receptor gene and protein expression and localization in human IUGR and normal term placentas
Mando C, et al.
Placenta, 32(1), 44-50 (2011)
Peina Wang et al.
Antioxidants (Basel, Switzerland), 11(7) (2022-07-28)
Blood-brain barrier (BBB) breakdown, a characteristic feature of ischemic stroke, contributes to poor patient outcomes. Brain microvascular endothelial cells (BMVECs) are a key component of the BBB and dysfunction or death of these cells following cerebral ischemia reperfusion (I/R) injury
Regulation of cellular iron metabolism
Wang J and Pantopoulos K
The Biochemical Journal, 434(3), 365-381 (2011)
Yidan Liang et al.
Brain research bulletin, 207, 110878-110878 (2024-01-14)
Cerebrovascular dysfunction resulting from traumatic brain injury (TBI) significantly contributes to poor patient outcomes. Recent studies revealed the involvement of iron metabolism in neuronal survival, yet its effect on vasculature remains unclear. This study aims to explore the impact of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service