SAB3500765
Anti-TLR4 antibody produced in rabbit
affinity isolated antibody
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
rabbit
Quality Level
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
mol wt
92 kDa
species reactivity
human
concentration
1 mg/mL
technique(s)
ELISA: suitable
immunohistochemistry: suitable
NCBI accession no.
UniProt accession no.
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... TLR4(7099)
General description
Toll-like receptor 4 (TLR4) belongs to the interleukin-1 receptor (IL-1R) family. It is highly polymorphic and is located on human chromosome 9q33.1.
Immunogen
Antibody was raised against a 14 amino acid peptide near the carboxy terminus of human TLR4.
Biochem/physiol Actions
Toll-like receptor 4 (TLR4) acts as the receptor for LPS (lipopolysaccharides ), a gram-negative bacterial cell wall component. Mutations in TLR4 affects mRNA and protein expression in colorectal cancer.
Features and Benefits
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.
Linkage
The action of this antibody can be blocked using blocking peptide SBP3500765.
Physical form
Supplied at 1 mg/mL in PBS with 0.02% sodium azide.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Not finding the right product?
Try our Product Selector Tool.
related product
Product No.
Description
Pricing
Storage Class Code
10 - Combustible liquids
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
TLR4 up-regulation at protein or gene level is pathogenic for lupus-like autoimmune disease.
Journal of Immunology, 177(10), 6880-6888 (2006)
Endotoxin and cytokine regulation of toll-like receptor (TLR) 2 and TLR4 gene expression in murine liver and hepatocytes.
Journal of Interferon & Cytokine Research, 20(10), 915-921 (2000)
TLR2 and TLR4 polymorphisms influence mRNA and protein expression in colorectal cancer.
World Journal of Gastroenterology, 21(25), 7730-7730 (2015)
International journal of biological sciences, 16(9), 1481-1494 (2020-04-01)
Inflammation and apoptosis are considered as two major pathological causes of human sarcopenia. The current understanding based on different models recognizes that apoptosis does not trigger inflammation, while emerging evidence indicates that inflammation can induce apoptosis. Here, we provide solid
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service