Skip to Content
Merck
All Photos(4)

Key Documents

SAB3500383

Sigma-Aldrich

Anti-CXCR4 antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

Anti-CXCR4

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, human

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunofluorescence: suitable
immunoprecipitation (IP): suitable
indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CXCR4(7852)

General description

C-X-C motif chemokine receptor 4 (CXCR4) is encoded by the gene mapped to human chromosome 2q22.1. The gene codes for a protein with seven transmembrane regions. The encoded protein is expressed on the surface of T-cells, B-cells, monocytes, neutrophils and dendritic cells.

Immunogen

CXCR4 antibody was raised against a peptide corresponding to amino acids near the amino terminus of human CXCR4.

Application

Anti-CXCR4 antibody produced in rabbit has been used in western blot analysis and indirect ELISA.
Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Western Blotting (1 paper)

Biochem/physiol Actions

C-X-C motif chemokine receptor 4 (CXCR4) is specific for stromal cell-derived factor-1. It plays a vital role in regulation of immune response in various immune cell types. CXCR4 interacts with CD4 (cluster of differentiation 4) and permits human immunodeficiency virus type 1 (HIV-1) entry and infection in to the cells. Polymorphism in the gene is associated with the development of Juvenile idiopathic arthritis (JIA). In addition, mutation in the gene increases the risk of susceptibility to cardiovascular diseases (CVDs).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500383.

Physical form

Supplied in PBS with 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CD4-independent infection by HIV-2 is mediated by fusin/CXCR4.
Endres M J, et al.
Cell, 87(4), 745-756 (1996)
Withaferin A targeting both cancer stem cells and metastatic cancer stem cells in the UP-LN1 carcinoma cell model.
T Lai-Lei, et al.
Journal of Cancer Metastasis and Treatment, 2(1), 29-40 (2016)
Deepika Nayak et al.
Journal of cell communication and signaling, 17(3), 609-626 (2022-11-04)
Cancer stem cells (CSCs) cause drug resistance in cancer due to its extensive drug efflux, DNA repair and self-renewal capability. ATP binding cassette subfamily G member 2 (ABCG2) efflux pump afford protection to CSCs in tumors, shielding them from the
Genetic variation of CXCR4 and risk of coronary artery disease: epidemiological study and functional validation of CRISPR/Cas9 system.
Runmin G, et al.
Oncotarget, 9(18), 14077?14083-14077?14083 (2018)
Ben Hitchinson et al.
Science signaling, 11(552) (2018-10-18)
Repeated dosing of drugs targeting G protein-coupled receptors can stimulate antagonist tolerance, which reduces their efficacy; thus, strategies to avoid tolerance are needed. The efficacy of AMD3100, a competitive antagonist of the chemokine receptor CXCR4 that mobilizes leukemic blasts from

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service