Skip to Content
Merck
All Photos(1)

Key Documents

SAB3500318

Sigma-Aldrich

Anti-TMEM16A antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Ano1, Anti-Anoctamin 1, Anti-Calcium activated chloride channel, Anti-DOG1, Anti-TAOS1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, human

technique(s)

indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ANO1(55107)

General description

Transmembrane member 16A (TMEM16A) or Anoctamin1 (Ano1) is a calcium-activated chloride channel. It is expressed in the interstitial cells of Cajal (ICC) in the gastrointestinal tract. The gene encoding this protein is localized on human chromosome 11q13.3.

Immunogen

TMEM16A antibody was raised against a 17 amino acid peptide from near the amino terminus of human TMEM16A.

Biochem/physiol Actions

Transmembrane member 16A (TMEM16A) or Ano1 is crucial for normal electrical slow waves and for the proliferation of interstitial cells of Cajal (ICC). The protein has a role in smooth muscle contraction, salivary secretion and cardiac and neuronal excitability. TMEM16A has been associated with head and neck squamous cell carcinoma and breast cancer.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500318.

Physical form

Supplied in PBS with 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

TMEM16A/ANO1 suppression improves response to antibody-mediated targeted therapy of EGFR and HER2/ERBB2.
Kulkarni S
Genes Chromosomes Cancer (2017)
EAVK segment "c" sequence confers Ca2+-dependent changes to the kinetics of full-length human Ano1.
Strege PR
American Journal of Physiology: Gastrointestinal and Liver Physiology (2017)
GDD1 Is Identical to TMEM16E, a Member of the TMEM16 Family
Masuko K
American Journal of Human Genetics (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service