Skip to Content
Merck
All Photos(1)

Key Documents

SAB2900695

Sigma-Aldrich

Anti-ADCYAP1R1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

ADCYAP1R1, PAC1, PAC1-R, PACAP receptor 1, Pac1 receptor, Pac1 receptors

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
IHC
clone:
polyclonal
species reactivity:
dog, human, Xenopus, chicken, monkey
citations:
2
technique(s):
immunohistochemistry: 15 μg/mL

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

dog, human, Xenopus, chicken, monkey

technique(s)

immunohistochemistry: 15 μg/mL

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ADCYAP1R1(117)

General description

Adenylate cyclase activating polypeptide 1 (pituitary) receptor type I (ADCYAP1R1) or pituitary adenylate cyclase 1 receptor (PAC1R) protein is a member of the class B G-protein-coupled receptors (GPCRs). It is expressed ubiquitously in the central nervous system (CNS) and peripheral tissues. It has an N-terminal extracellular domain (ECD) and a transmembrane domain (TMD). ADCYAP1R1 gene is located on human chromosome 7p14.

Immunogen

Synthetic 17 amino acid peptide from C-terminus of human ADCYAP1R1. Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Panda, Dog, Bat, Chicken, Lizard, Xenopus (100%); Stickleback, Pufferfish, Zebrafish (94%); Mouse, Rat, Hamster, Elephant, Bovine, Horse, Pig (88%).

Biochem/physiol Actions

Adenylate cyclase activating polypeptide 1 (pituitary) receptor type I (ADCYAP1R1) or pituitary adenylate cyclase 1 receptor (PAC1R) protein can activate the adenylyl cyclase stimulatory G protein Gs. It regulates circadian rhythm, food intake, and the immune system. PAC1R plays a vital role in glucose metabolism, learning and memory, neuronal ontogenesis, and apoptosis. ADCYAP1R1 is identified as a potential drug target for mental disorders and dry eye syndrome.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate-buffered saline containing less than 0.1% sodium azide.
Supplied at 1 mg/ml

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kazuhiro Kobayashi et al.
Nature structural & molecular biology, 27(3), 274-280 (2020-03-12)
Pituitary adenylate cyclase-activating polypeptide (PACAP) is a pleiotropic neuropeptide hormone. The PACAP receptor PAC1R, which belongs to the class B G-protein-coupled receptors (GPCRs), is a drug target for mental disorders and dry eye syndrome. Here, we present a cryo-EM structure
M Stoffel et al.
Genomics, 23(3), 697-699 (1994-10-01)
The gene encoding the human type I pituitary adenylate cyclase activating polypeptide receptor (ADCYAP1R1) was mapped to chromosome 7 by PCR analysis of genomic DNA from a human/rodent somatic cell hybrid mapping panel. This assignment was confirmed and the gene

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service