Skip to Content
Merck
All Photos(4)

Key Documents

SAB2500431

Sigma-Aldrich

Anti-FTL antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Ferritin L subunit, Anti-L apoferritin, Anti-MGC71996

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

rat, mouse, human

technique(s)

immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FTL(2512)

General description

Ferritin light (FTL) subunit along with ferritin heavy (FTH) subunit are members of the ferritin complex. FTL gene is mapped on the human chromosome at 19q13.33.

Immunogen

Peptide with sequence C-GEYLFERLTLKHD from the C Terminus of the protein sequence according to NP_000137.2.

Application

Anti-FTL antibody produced in goat has been used in western blotting (1:500).

Biochem/physiol Actions

Ferritin light (FTL) subunit is an important protein in iron metabolism but lacks catalytic activity. This protein participates in the nucleation and mineralization of the iron center. FTL acts as a biomarker to differentiate between malignant and benign tumors. Overexpression of the FTL gene is observed in several malignant tumors and plays an important role in mediating the malignancy of cancers. FTL gene plays a role in the pathogenesis of glioblastoma. Mutation or deletion of the FTL gene is associated with neuroferritinopathy.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mimi C Sammarco et al.
The Journal of biological chemistry, 283(8), 4578-4587 (2007-12-28)
Ferritin plays an important role in the storage and release of iron, an element utilized in cellular processes such as respiration, gene regulation, and DNA replication and repair. Ferritin in animals is composed of 24 ferritin L (FTL) and ferritin
Junhui Liu et al.
Journal of experimental & clinical cancer research : CR, 39(1), 137-137 (2020-07-18)
Hypoxia, a fundamental characteristic of glioma, is considered to promote tumor malignancy by inducing process of epithelial mesenchymal transition (EMT). Ferritin Light Chain (FTL) is one of the iron metabolism regulators and is overexpressed in glioma. However, relationship between hypoxia
Sara Zumerle et al.
Blood, 123(23), 3646-3650 (2014-03-22)
Hepcidin is a 25-amino-acid peptide demonstrated to be the iron regulatory hormone capable of blocking iron absorption from the duodenum and iron release from macrophages. Mutations affecting hepcidin regulators or the hepcidin gene itself cause hemochromatosis, a common genetic disorder.
Jean-Christophe Deschemin et al.
Frontiers in physiology, 8, 804-804 (2017-11-02)
Pulmonary iron excess is deleterious and contributes to a range of chronic and acute inflammatory diseases. Optimal lung iron concentration is maintained through dynamic regulation of iron transport and storage proteins. The iron-regulatory hormone hepcidin is also expressed in the
Shinsuke Fujioka et al.
Neuro-degenerative diseases, 10(1-4), 257-260 (2012-01-21)
Major progress in genetic studies of Parkinson's disease (PD) and parkinsonism has been achieved in the last two decades. We provide a brief review of the current status of PARK and non-PARK loci/genes, and discuss two new genes: eIF4G1 and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service