Skip to Content
Merck
All Photos(1)

Documents

SAB2108661

Sigma-Aldrich

Anti-FFAR1

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

31 kDa

species reactivity

bovine, human, dog

concentration

0.5-1 mg/mL

technique(s)

immunoblotting: suitable

accession no.

NM_005303

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... FFAR1(2864)

General description

Free fatty acid receptor 1 (FFAR1) formerly known as G-protein coupled receptor 40 (GPR 40) is predominantly expressed in pancreatic beta cells. In human chromosome, the gene FFAR1 is located on 19q13.12.

Immunogen

Synthetic peptide directed towards the N terminal region of human FFAR1

Biochem/physiol Actions

Free fatty acid receptor 1 (FFAR1) is activated by binding of medium or long chain free fatty acids. It increases the Ca2+ level intracellularly through phospholipase C mediated signalling and that potentially stimulates insulin production. FFAR1 is a potential drug target for metabolic diseases like type 2 diabetes.

Sequence

Synthetic peptide located within the following region: KVSEAALSLLVLILIITSASRSQPKVPEWVNTPSTCCLKYYEKVLPRRLV

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Free fatty acids regulate insulin secretion from pancreatic beta cells through GPR40
Itoh Y, et al.
Nature, 422(6928), 173-176 (2003)
Free fatty acid receptors FFAR1 and GPR120 as novel therapeutic targets for metabolic disorders
Hara T, et al.
Journal of Pharmaceutical Sciences, 100(9), 3594-3601 (2011)
Re-evaluation of fatty acid receptor 1 (FFAR1/GPR40) as drug target for the stimulation of insulin secretion in humans
Wagner R, et al.
Diabetes, DB_121249-DB_121249 (2013)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service